Skip to Content
Merck
All Photos(3)

Key Documents

HPA019044

Sigma-Aldrich

Anti-CFLAR antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-FLIP, Anti-CASH, Anti-CASP8 and FADD-like apoptosis regulator, Anti-CASP8AP1, Anti-CLARP, Anti-Casper, Anti-FLAME, Anti-FLIP, Anti-I-FLICE, Anti-MRIT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

IHQVEEALDTDEKEMLLFLCRDVAIDVVPPNVRDLLDILRERGKLSVGDLAELLYRVRRFDLLKRILKMDRKAVETHLLRNPHLVSDYRVLMAEIGEDLDKSDVSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFLAR(8837)

General description

The gene CFLAR (CASP8 and FADD-like apoptosis regulator) is mapped to human chromosome 2q33-q34. The protein localizes in the cytoplasm. CFLAR is commonly referred as c-FLIP (Cellular FLICE-like inhibitory protein).

Immunogen

CASP8 and FADD-like apoptosis regulator recombinant protein epitope signature tag (PrEST)

Application

Anti-CFLAR antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CFLAR (CASP8 and FADD-like apoptosis regulator) has been shown to suppress CD95-mediated apoptosis via inhibition of caspase-8 processing at the death-inducing signaling complex (DISC). Increase in levels of CFLAR inhibits Hepatitis B virus (HBV) X protein-mediated apoptosis. Presence of CFLAR protects cells from Fas, TNF (tumor necrosis factor) and TRAIL (TNF-related apoptosis-inducing ligand) receptors induced apoptosis. CFLAR is also associated with tumor growth and escape from immunity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74820

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Stacey Ricci et al.
Molecular and cellular biology, 24(19), 8541-8555 (2004-09-16)
Tumor necrosis factor alpha (TNF-alpha)-related apoptosis-inducing ligand (TRAIL) is a member of the TNF-alpha family of death receptor ligands and holds great therapeutic potential as a tumor cell-specific cytotoxic agent. Using a panel of established tumor cell lines and normal
Kyun-Hwan Kim et al.
The EMBO journal, 22(9), 2104-2116 (2003-05-03)
Despite its implication in the progression of hepatitis B virus (HBV)-associated liver disease, the pro-apoptotic function of HBx protein remains poorly understood. We show that the expression of HBx leads to hyperactivation of caspase-8 and caspase-3 upon treatment with tumor
C Scaffidi et al.
The Journal of biological chemistry, 274(3), 1541-1548 (1999-01-09)
Upon stimulation, CD95 (APO-1/Fas) recruits the adapter molecule Fas-associated death domain protein (FADD)/MORT1 and caspase-8 (FADD-like interleukin-1beta-converting enzyme (FLICE)/MACH/MCH5) into the death-inducing signaling complex (DISC). Recently, a molecule with sequence homology to caspase-8 was identified, termed cellular FLICE-inhibitory protein (c-FLIP).
David R Jones et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 7(11), 1683-1690 (2012-10-13)
Despite complete surgical resection survival in early-stage non-small-cell lung cancer (NSCLC) remains poor. On the basis of prior preclinical evaluations, we hypothesized that combined induction proteasome and histone deacetylase inhibitor therapy, followed by tumor resection, is feasible. A phase I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service