Skip to Content
Merck
All Photos(2)

Key Documents

HPA018191

Sigma-Aldrich

Anti-SESN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hi95, Anti-Sestrin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

HSLSSFVFGCGILPEGDADGSPAPQAPTPPSEQSSPPSRDPLNNSGGFESARDVEALMERMQQLQESLLRDEGTSQEEMESRFELEKSESLLVTPSADILEPSPHPDMLCFVEDPTFGY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SESN2(83667)

General description

The gene sestrin-2 (SENS2) is mapped to human chromosome 1p35.3. SENS2 belongs to sestrin family of conserved antioxidant proteins.

Immunogen

Sestrin-2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In mammalian cells, sestrin-2 (SENS2) decreases oxidative stress by facilitating the peroxidase activity of over-oxidised peroxiredoxins. SENS2 is highly expressed in the emphysematous lungs of humans with chronic obstructive pulmonary disease and lungs of habitual smokers. Mutational inactivation of SENS2 in mice prevents the development of cigarette-smoke-induced pulmonary emphysema by inducing an increase in PDGFRβ (platelet-derived growth factor receptor β) signaling. Absence of SENS2 prepares squamous cell carcinoma cells to 5-fluorouracil-induced apoptosis and melanoma cells to UV (ultraviolet)B- and vemurafenib-induced apoptosis, suggesting oncogenic role of SENS2. Overexpression of SENS2 in adipose tissues inhibits uncoupling protein-1 expression, resulting in decreased thermogenesis and increased fat accumulation.
Stimulation of dopamine D2 receptor increase endogenous renal SENS2 expression which protects against oxidative stress and is involved in the maintenance of normal blood pressure. Similarly, c-Jun NH2-terminal kinase (JNK)/c-Jun pathway increases SENS2 activity, protecting against vascular endothelial damage induced by angiotensin II.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72800

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lao Yi et al.
Cell biology and toxicology, 30(3), 147-156 (2014-05-20)
Sestrin-2 (SESN2) is involved in the cellular response to different stress conditions. However, the function of SESN2 in the cardiovascular system remains unknown. In the present study, we tested whether SESN2 has a beneficial effect on vascular endothelial damage induced
Seung-Hyun Ro et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(21), 7849-7854 (2014-05-16)
Uncoupling protein 1 (Ucp1), which is localized in the mitochondrial inner membrane of mammalian brown adipose tissue (BAT), generates heat by uncoupling oxidative phosphorylation. Upon cold exposure or nutritional abundance, sympathetic neurons stimulate BAT to express Ucp1 to induce energy
Baozhong Zhao et al.
The Journal of biological chemistry, 289(52), 35806-35814 (2014-11-08)
Skin cancer is the most common cancer in the United States and is mainly caused by environmental UV radiation. Reducing skin cancer incidence is becoming an urgent issue. The stress-inducible protein Sestrin2 (Sesn2) plays an important role in maintaining redox
Yu Yang et al.
Hypertension (Dallas, Tex. : 1979), 64(4), 825-832 (2014-07-16)
The dopamine D2 receptor (D2R) decreases renal reactive oxygen species (ROS) production and regulates blood pressure, in part, via positive regulation of paraoxonase 2. Sestrin2, a highly conserved antioxidant protein, regulates intracellular ROS level by regenerating hyperoxidized peroxiredoxins. We hypothesized
Regine Bergholdt et al.
PloS one, 4(7), e6250-e6250 (2009-07-18)
Proteins contributing to a complex disease are often members of the same functional pathways. Elucidation of such pathways may provide increased knowledge about functional mechanisms underlying disease. By combining genetic interactions in Type 1 Diabetes (T1D) with protein interaction data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service