Skip to Content
Merck
All Photos(7)

Key Documents

HPA017139

Sigma-Aldrich

Anti-CD276 antibody produced in rabbit

enhanced validation

affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution

Synonym(s):

Anti-4Ig-B7-H3 antibody produced in rabbit, Anti-B7 homolog 3 antibody produced in rabbit, Anti-B7-H3 antibody produced in rabbit, Anti-CD276 antigen precursor antibody produced in rabbit, Anti-Costimulatory molecule antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EVQVPEDPVVALVGTDATLCCSFSPEPGFSLAQLNLIWQLTDTKQLVHSFAEGQDQGSAYANRTALFPDLLAQGNASLRLQRVRVADEGSFTCFVSIRDFG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD276(80381)

Looking for similar products? Visit Product Comparison Guide

General description

CD276 is a costimulatory molecule belonging to the B7 family. It is expressed in various normal tissues and in several tumor cell lines.

Immunogen

CD276 antigen precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CD276 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CD276 is involved in the T cell activation and IFN (interferon)-γ production. It directly binds to putative counter-receptor on activated T cells to stimulate proliferation of both CD4+ and CD8+ T cells. It has been reported that CD276 may work as a biomarker in the pathogenesis of rheumatoid arthritis. CD276 is associated with the signal transducer and activator of transcription 3 (STAT3) signaling pathway in the human hepatocellular carcinoma (HCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71719

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dieter Lemke et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(1), 105-117 (2011-11-15)
Recent work points out a role of B7H3, a member of the B7-family of costimulatory proteins, in conveying immunosuppression and enforced invasiveness in a variety of tumor entities. Glioblastoma is armed with effective immunosuppressive properties resulting in an impaired recognition
Jing Sun et al.
Clinical immunology (Orlando, Fla.), 159(1), 23-32 (2015-05-02)
CD276 (B7-H3) is a costimulatory molecule that plays a potent role in T cell responses, however, the role of B7-H3 in autoimmune diseases has not been elucidated. We analyzed B7-H3 expression in rheumatoid arthritis (RA) for the first time and
A I Chapoval et al.
Nature immunology, 2(3), 269-274 (2001-02-27)
We describe here a newly identified member of the human B7 family, designated B7 homolog 3 (B7-H3), that shares 20-27% amino acid identity with other B7 family members. B7-H3 mRNA is not detectable in peripheral blood mononuclear cells, although it
Fu-Biao Kang et al.
Oncology reports, 33(1), 274-282 (2014-11-06)
B7 family members are aberrantly expressed on the human hepatocellular carcinoma (HCC) cell surface, and induce local and systemic immunosuppression. Tumor-associated macrophages (TAMs) are a significant immune cell subpopulation in HCC and may be induced to express co-inhibitory molecules including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service