Skip to Content
Merck
All Photos(7)

Key Documents

HPA011078

Sigma-Aldrich

Anti-HLA-DPB1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HLA class II histocompatibility antigen, DP(W4) β-chain precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

YNREEFVRFDSDVGEFRAVTELGRPDEEYWNSQKDILEEKRAVPDRMCRHNYELGGPMTLQRRVQPRVNVSPSKKGPLQHHNLLVCHVTDFYPGSIQVRWFLNGQEETAGVVSTNLIRNGDWTF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HLA-DPB1(3115)

General description

HLA-DPB1 (major histocompatibility complex, class II, DP β 1) belongs to HLA (human leukocyte antigen) class II family of molecules, which are heterodimers of α and β chains. It is present on human chromosome 6p21.3. It is a highly polymorphic glycoprotein which is present on the surface of antigen presenting cells (APCs). The α and β chains of HLA-DPB1 have two extracellular domains, a hydrophobic transmembrane region and a hydrophilic cytoplasmic tail. It has two α and β chain types- DPα1 and DPα2, and DPβ1 and DPβ2, where DPα2 and DPβ2 are pseudogenes.

Immunogen

HLA class II histocompatibility antigen, DP(W4) β-chain precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HLA-DPB1 (major histocompatibility complex, class II, DP β 1) recognizes and presents antigens to T-helper cells. Polymorphisms in this gene are linked to the differences and level of response to rubella vaccination. Variants of this gene are associated with systemic sclerosis, where DPB1*03:01 and *13:01 alleles are up-regulated. Patients with DPB1*03:01 allele also had higher chances of developing pulmonary fibrosis. HLA-DPB1 05:01 allele is associated with long-term memory against hepatitis B surface antigen (HBsAg), following hepatitis B vaccination. Mismatch in HLA-DPB1 allele during hematopoietic cell transplantation, from an unrelated donor, increases the risk acute graft-versus-host disease (aGVHD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72202

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiucun Wang et al.
PloS one, 9(1), e87363-e87363 (2014-02-06)
Human leukocyte antigen DPB1 was reported to contain singly nucleotide polymorphisms conferring the strongest susceptibility to systemic sclerosis in Korean population. However, associations of specific DPB1 alleles with SSc vary in different ethnic populations. The aim of this study was
T-W Wu et al.
Genes and immunity, 15(1), 47-53 (2013-11-29)
Previously we reported significant associations of the human leukocyte antigen (HLA)-DPB1 05:01 with memory against hepatitis B (HB) vaccination. However, the effects of HLA-DPB1 on antibodies to hepatitis B surface antigen (anti-HBs) kinetics were not explored. We followed up a
E Maruya et al.
Genome research, 6(1), 51-57 (1996-01-01)
We have developed a simple and efficient procedure with which to form single-stranded DNA [ssDNA] and then applied HLA-DRB1, -DQB1, and -DPB1 allele typing. This method is referred to as low ionic strength single-stranded conformation polymorphism (LIS-SSCP), and is based
Seisuke Kusano et al.
Journal of molecular biology, 426(17), 3016-3027 (2014-07-16)
The major allergen, Cry j 1, was isolated from Japanese cedar Cryptomeria japonica (Cry j) pollen and was shown to react with immunoglobulin E antibodies in the sera from pollinosis patients. We previously reported that the frequency of HLA-DP5 was
Bronwen E Shaw et al.
Blood, 110(13), 4560-4566 (2007-08-30)
Hematopoietic cell transplantation (HCT) from an HLA-A, HLA-B, HLA-C, HLA-DRB1, and HLA-DQB1 allele-matched unrelated donor is a well-recognized life-saving treatment modality for patients with hematologic disorders. The morbidity and mortality from clinically significant acute graft-versus-host disease (aGVHD) remains a limitation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service