Skip to Content
Merck
All Photos(5)

Key Documents

HPA007105

Sigma-Aldrich

Anti-ALPL antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AP-TNAP antibody produced in rabbit, Anti-Alkaline phosphatase liver/bone/kidney isozyme antibody produced in rabbit, Anti-Alkaline phosphatase, tissue-nonspecific isozyme precursor antibody produced in rabbit, Anti-TNSALP antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LTSSEDTLTVVTADHSHVFTFGGYTPRGNSIFGLAPMLSDTDKKPFTAILYGNGPGYKVVGGERENVSMVDYAHNNYQAQSAVPLRHETHGGEDVAVFSKGPMAHLLHGVHEQNYVPHVMAYAACIGANL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALPL(249)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Alkaline phosphatase, tissue-nonspecific isozyme precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ALPL (Alkaline phosphatase) is a marker enzyme, expressed in the brush border membrane of proximal tubular cells. It encodes a tissue-non-specific alkaline phosphatase isoenzyme. It plays a major role in the cell viability and apoptosis. Thus, ALPL has been proposed as an effective candidate for tumorigenesis. Mutation in the ALPL gene activity causes a rare inherited disorder, hypophosphatasia, characterized by defective bone mineralization and deficiency of serum.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71397

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiou Yang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 32(3), 635-644 (2013-09-12)
Hypophosphatasia, a rare inherited disease characterized by defective mineralization of bone and teeth, is caused by various mutations in the tissue-nonspecific isoenzyme of alkaline phosphatase (TNSALP) gene. Our aim was to determine the mutations on TNSALP gene in three Chinese
Ujjawal Sharma et al.
Biochimie, 104, 27-35 (2014-06-10)
Renal cell carcinoma (RCC) is the most common kidney cancer in adults. Although several genes have been found to be involved in carcinogenesis of RCC, more great efforts are needed to identify new genes which are responsible for the process.
Takeshi Taketani et al.
Archives of disease in childhood, 99(3), 211-215 (2013-11-28)
We examined the clinical and genetic features of hypophosphatasia (HPP) in Japanese patients. HPP is a rare metabolic bone disorder of bone mineralisation caused by mutations in the liver/bone/kidney alkaline phosphatase (ALPL) gene, which encodes tissue-non-specific alkaline phosphatase isoenzyme. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service