Skip to Content
Merck
All Photos(3)

Key Documents

HPA006294

Sigma-Aldrich

Anti-LZTS1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-F37/esophageal cancer-related gene-coding leucine-zipper motif antibody produced in rabbit, Anti-Fez1 antibody produced in rabbit, Anti-Leucine zipper putative tumor suppressor 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TEVNAKASEILGLKAQLKDTRGKLEGLELRTQDLEGALRTKGLELEVCENELQRKKNEAELLREKVNLLEQELQELRAQAALARDMGPPTFPEDVPALQRELERLRAELREERQGHDQMSSGFQHERLVWKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LZTS1(11178)

Immunogen

Leucine zipper putative tumor suppressor 1 recombinant protein epitope signature tag (PrEST)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70846

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axelle Wilmerding et al.
Development (Cambridge, England), 148(4) (2021-01-22)
Differential Hox gene expression is central for specification of axial neuronal diversity in the spinal cord. Here, we uncover an additional function of Hox proteins in the developing spinal cord, restricted to B cluster Hox genes. We found that members
Francesca Lovat et al.
Oncotarget, 5(4), 970-977 (2014-01-23)
The Leucine Zipper Tumor Suppressor 1 (LZTS1) is a tumor suppressor gene, located at chromosome 8p22, which is frequently altered in human cancer. In normal tissue, its ubiquitous expression regulates cell mitosis by the stabilization of microtubule networks. LZTS1-deficient mouse
Marlene Kropp et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 241(5), 984-994 (2012-03-16)
Neuronal circuit assembly comprises a number of developmental processes that ultimately underlie function. Identifying the molecular events that dictate these processes can give key insights into how neuronal circuit formation is coordinated. To begin to identify such molecular mechanisms, we
T Kawaue et al.
Nature communications, 10(1), 2780-2780 (2019-06-27)
In the developing central nervous system, cell departure from the apical surface is the initial and fundamental step to form the 3D, organized architecture. Both delamination of differentiating cells and repositioning of progenitors to generate outer radial glial cells (oRGs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service