Skip to Content
Merck
All Photos(9)

Key Documents

HPA004765

Sigma-Aldrich

Anti-NGFR antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD271 antigen antibody produced in rabbit, Anti-Gp80-LNGFR antibody produced in rabbit, Anti-Low affinity neurotrophin receptor p75NTR antibody produced in rabbit, Anti-Low-affinity nerve growth factor receptor antibody produced in rabbit, Anti-NGF receptor antibody produced in rabbit, Anti-Tumor necrosis factor receptor superfamily member 16 precursor antibody produced in rabbit, Anti-p75 ICD antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NKQGANSRPVNQTPPPEGEKLHSDSGISVDSQSLHDQQPHTQTASGQALKGDGGLYSSLPPAKREEVEKLLNGSAGDTWRHLAGELGYQPEHIDSFTHEACPVRALLASWATQDSATLDALLAALRRIQRADLVESLCSES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)

General description

NGFR (nerve growth factor receptor) is a cell surface transmembrane glycoprotein. It belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. It is expressed on the surface of glial fibrillary acidic protein δ (GFAPδ) cells of the human brain. In addition to the brain, it is also expressed in liver, lung, and muscle cells.

Immunogen

Tumor necrosis factor receptor superfamily member 16 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NGFR (nerve growth factor receptor) plays a crucial role in the neuronal survival, differentiation and development of neurons in adult brain. It has been reported that the membrane-spanning region and cytoplasmic domain of NGFR plays an essential role in signal transduction during neuronal differentiation. In CNS and peripheral tissues, NGFR acts as a clock gene and its activity is modulated by helix-loop-helix transcription factors: CLOCK and BMAL1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70014

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan C Johnson et al.
American journal of clinical pathology, 142(6), 795-802 (2014-11-13)
We retrospectively tested the prognostic and diagnostic significance of CD271+ mesenchymal stromal cell (MSC) density in cytopenic patients who underwent bone marrow biopsy to evaluate for myelodysplastic syndromes (MDS). CD271+ MSC density was quantitated by automated image analysis of tissue
D L Baker et al.
The American journal of pathology, 139(1), 115-122 (1991-07-01)
Nerve growth factor (NGF) is important to the survival, development, and differentiation of neurons. Its action is mediated by a specific cell surface transmembrane glycoprotein, nerve growth factor receptor (NGFR). In this study, NGFR expression by human fetal and adult
Miriam E van Strien et al.
Stem cells translational medicine, 3(4), 470-480 (2014-03-08)
Neural progenitor cells (NPCs) in the subventricular zone (SVZ) hold promise for future therapy for neurodegenerative disorders, because the stimulation of adult neurogenesis could potentially restore the function of degenerating neurons and glia. To obtain more knowledge on these NPCs
H Yan et al.
Science (New York, N.Y.), 252(5005), 561-563 (1991-04-26)
To determine the domains of the low-affinity nerve growth factor (NGF) receptor required for appropriate signal transduction, a series of hybrid receptors were constructed that consisted of the extracellular ligand-binding domain of the human epidermal growth factor (EGF) receptor (EGFR)
Kazunari Aoki et al.
British journal of haematology, 193(3), 659-668 (2021-04-11)
A population of mesenchymal stem cells, termed CXC chemokine ligand (CXCL)12-abundant reticular (CAR) cells or leptin receptor-expressing cells, are the major cellular component of niches for haematopoietic stem cells (HSCs) in murine bone marrow. CAR cells are characterized by several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service