Skip to Content
Merck
All Photos(7)

Key Documents

HPA003317

Sigma-Aldrich

Anti-CUX1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CASP, Anti-CDP, Anti-CDP/Cut, Anti-CDP/Cux, Anti-CDP1, Anti-CUT, Anti-CUTL1, Anti-CUX, Anti-Clox, Anti-Cux/CDP, Anti-GOLIM6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KSFQGEIDALSKRSKEAEAAFLNVYKRLIDVPDPVPALDLGQQLQLKVQRLHDIETENQKLRETLEEYNKEFAEVKNQEVTIKALKEKIREYEQTLKNQAETIALEKEQKLQNDFAEKERKLQETQMSTTSKLEEAEHKVQSLQTALE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUX1(1523)

Immunogen

cut-like homeobox 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CUX1 (cut-like homeobox 1) gene encodes a transcription factor belonging to the homeodomain family of DNA binding proteins. It functions as a transcriptional repressor and as an activator regulating cell differentiation and cell cycle progression. It modulates the invasion in various cancers and mediates resistance to apoptosis. It is involved in tumor cell survival in pancreatic cancer. It is involved in the control of food intake in both rodents and humans by regulating the expression of the FTO (Fat Mass and Obesity Associated) and RPGRIP1L (Retinitis Pigmentosa GTPase Regulator-interacting Protein-1-like) genes. It also coordinates leptin receptor signaling. Cux1 and Cux2 are involved in the regulation of dendrite branching, spine development, and synapse formation in layer II-III neurons of the cerebral cortex.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84785

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pinar Mesci et al.
PLoS biology, 20(11), e3001845-e3001845 (2022-11-04)
The Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) is the causative agent of coronavirus disease 2019 (COVID-19), which was rapidly declared a pandemic by the World Health Organization (WHO). Early clinical symptomatology focused mainly on respiratory illnesses. However, a variety
Beatriz Cubelos et al.
Neuron, 66(4), 523-535 (2010-06-01)
Dendrite branching and spine formation determines the function of morphologically distinct and specialized neuronal subclasses. However, little is known about the programs instructing specific branching patterns in vertebrate neurons and whether such programs influence dendritic spines and synapses. Using knockout
S Ripka et al.
Gut, 59(8), 1101-1110 (2010-05-06)
The transcription factor CUX1 is known as a regulator of cell differentiation and cell cycle progression. Previously, CUX1 was identified as a modulator of invasiveness in various cancers. Based on expression profiles suggesting a role for CUX1 in mediating chemoresistance
Marta Nieto et al.
The Journal of comparative neurology, 479(2), 168-180 (2004-09-29)
Little is known about how neurons in the different layers of the mammalian cerebral cortex are specified at the molecular level. Expression of two homologues of the Drosophila homeobox Cut gene, Cux-1 and Cux-2, is strikingly specific to the pyramidal
Stefanie Ripka et al.
Neoplasia (New York, N.Y.), 12(8), 659-667 (2010-08-07)
Previously, we identified the transcription factor CUX1 as an important modulator of invasion and resistance to apoptosis. Expression profiles suggested that CUX1 regulates a complex transcriptional program mediating tumor progression. We aimed to identify functionally relevant targets of CUX1 by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service