Skip to Content
Merck
All Photos(6)

Key Documents

HPA002822

Sigma-Aldrich

Anti-EMILIN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EMILIN-1 precursor antibody produced in rabbit, Anti-Elastin microfibril interface-located protein 1 antibody produced in rabbit, Anti-Elastin microfibril interfacer 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PQSIMYRRFLRPRYRVAYKTVTDMEWRCCQGYGGDDCAESPAPALGPASSTPRPLARPARPNLSGSSAGSPLSGLGGEGPGESEKVQQLEEQVQSLTKELQGLRGVLQGLSGRLAEDVQRAVETAFNGRQQPADAAARPGVHETLNE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EMILIN1(11117)

Immunogen

EMILIN-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-EMILIN1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

The gene EMILIN1 encodes elastin microfibril interfacer-1 protein that contributes to the pathogenesis of essential hypertension (EH) in humans. It maintains elastic fibres in vessel walls and peripheral arterial compliance. It is considered as a candidate gene for human EH. It may regulate the formation of oxytalan fibers and play a role in their homeostasis. It acts as a negative regulator of the transforming growth factor-beta (TGF-β) signaling, which is involved in blood pressure (BP) homeostasis. The gene participates in the transcription of elastogenesis of elastic extracellular matrix (ECM′s) formation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85200

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chikako Kanno Honda et al.
Theranostics, 14(5), 1873-1885 (2024-03-20)
Rationale: The tumor microenvironment (TME) and its multifaceted interactions with cancer cells are major targets for cancer treatment. Single-cell technologies have brought major insights into the TME, but the resulting complexity often precludes conclusions on function. Methods: We combined single-cell
V M S Oh et al.
Journal of human hypertension, 26(9), 553-561 (2011-07-15)
Studies in mice suggest that the elastin microfibril interfacer-1 gene (EMILIN1), the gene encoding elastin microfibril interfacer-1 protein, contributes to the pathogenesis of essential hypertension (EH) in humans. EMILIN1 in part maintains elastic fibres in vessel walls, and hence peripheral
Yuka Nakatomi et al.
Connective tissue research, 52(1), 30-35 (2010-08-13)
The elastic system fibers comprise oxytalan, elaunin, and elastic fibers, differing in their relative microfibril and elastin contents. Among them, human periodontal ligament (PDL) contains only oxytalan fibers (pure microfibrils). Elastin microfibril interface-located protein-1 (EMILIN-1) is localized at the interface
Shinya Kondo et al.
Journal of cosmetic dermatology, 21(10), 4796-4804 (2022-01-23)
The elastic fiber structure becomes shorter, thicker, and curved with age. Nonetheless, the proteins and catabolic enzymes influencing the maintenance of and change in the three-dimensional (3D) structure of elastic fibers remain unknown. This study aimed to identify the proteins
Laura Louzao-Martinez et al.
Matrix biology plus, 4, 100011-100011 (2019-07-25)
Cell-based approaches using tissue engineering and regenerative medicine to replace damaged renal tissue with 3D constructs is a promising emerging therapy for kidney disease. Besides living cells, a template provided by a scaffold based on biomaterials and bioactive factors is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service