Skip to Content
Merck
All Photos(6)

Key Documents

HPA001934

Sigma-Aldrich

Anti-FBLN2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fibulin-2 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLN2(2199)

General description

Fibulin-2 (FBLN2) is an extracellular matrix protein, expressed predominantly in the heart and skin. It has been reported as a candidate gene for Marfan syndrome type 2.

Immunogen

Fibulin-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FBLN2 (fibulin 2) forms a linkage to connect laminin-1 and laminin-5 with other extracellular matrix proteins. By linking two of them FBLN2 establish communication between the cell surface and the basement membrane. It plays an important role in establishing laminin network.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86478

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Schaeffer et al.
Developmental biology, 442(1), 101-114 (2018-06-27)
During amniote peripheral nervous system development, segmentation ensures the correct patterning of the spinal nerves relative to the vertebral column. Along the antero-posterior (rostro-caudal) axis, each somite-derived posterior half-sclerotome expresses repellent molecules to restrict axon growth and neural crest migration
G Collod et al.
European journal of human genetics : EJHG, 4(5), 292-295 (1996-01-01)
Fibulin-2 (FBLN2) is a new extracellular matrix protein that has been considered a candidate gene for Marfan syndrome type 2 (locus MFS2) based on chromosomal colocation at 3p24.2-p25 and disease phenotype. In the absence of polymorphic markers reported for FBLN2
A K Langton et al.
The British journal of dermatology, 177(3), 818-827 (2017-01-31)
Human skin has the crucial roles of maintaining homeostasis and protecting against the external environment. Skin offers protection against mechanical trauma due to the reversible deformation of its structure; these biomechanical properties are amenable to dynamic testing using noninvasive devices.
H Kuivaniemi et al.
European journal of human genetics : EJHG, 6(6), 642-646 (1999-01-15)
We used conformation sensitive gel electrophoresis and direct sequencing of PCR products to screen for mutations in the cDNA for fibulin-2, an extracellular matrix protein, from 11 patients with abdominal aortic aneurysms and two controls. When compared with the published
A Utani et al.
The Journal of biological chemistry, 272(5), 2814-2820 (1997-01-31)
Epithelial cell-specific laminin-5, consisting of three chains, alpha3, beta3, and gamma2, is a component of the anchoring filament that traverses the lamina lucida beneath the hemidesmosomes of epidermal cells and functions to link these cells to the basement membrane. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service