Skip to Content
Merck
All Photos(4)

Key Documents

HPA001566

Sigma-Aldrich

Anti-CYB5R3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-B5R antibody produced in rabbit, Anti-Cytochrome b5 reductase 3 antibody produced in rabbit, Anti-Diaphorase-1 antibody produced in rabbit, Anti-NADH-cytochrome b5 reductase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

SGLLVYQGKGKFAIRPDKKSNPIIRTVKSVGMIAGGTGITPMLQVIRAIMKDPDDHTVCHLLFANQTEKDILLRPELEELRNKHSARFKLWYTLDRAPEAWDYGQGFVNEEMIRDHLPPPEEEPLVLMCGPPPMIQYACLPNLDH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYB5R3(1727)

Related Categories

Immunogen

NADH-cytochrome b5 reductase recombinant protein epitope signature tag (PrEST)

Application

Anti-CYB5R3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

CYB5R3 (cytochrome b5 reductase 3) gene encodes cytochrome b5 reductase that may be either in the membrane-bound form in somatic cell or in the soluble form in erythrocytes. The membrane-bound form is found anchored to the ER and is involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, and in drug metabolism. It has both membrane-binding and catalytic domains. The soluble form has only the catalytic domain and functions in methemoglobin reduction. It is involved in the regulation of NAD+/NADH ratio, in turn affecting aerobic metabolism and aging processes. Defects in this gene cause methemoglobinemias.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73379

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael de Cabo et al.
Aging, 2(1), 63-68 (2009-01-01)
Aging results from a complex and not completely understood chain of processes that are associated with various negative metabolic consequences and ultimately leads to senescence and death. The intracellular ratio of pyridine nucleotides (NAD(+)/NADH), has been proposed to be at
Heyka H Jakobs et al.
PloS one, 9(8), e105371-e105371 (2014-08-22)
The mitochondrial amidoxime reducing component mARC is the fourth mammalian molybdenum enzyme. The protein is capable of reducing N-oxygenated structures, but requires cytochrome b5 and cytochrome b5 reductase for electron transfer to catalyze such reactions. It is well accepted that
Sophia Rixen et al.
The Journal of biological chemistry, 294(46), 17593-17602 (2019-09-27)
The mitochondrial amidoxime-reducing component (MARC) is a mammalian molybdenum-containing enzyme. All annotated mammalian genomes harbor two MARC genes, MARC1 and MARC2, which share a high degree of sequence similarity. Both molybdoenzymes reduce a variety of N-hydroxylated compounds. Besides their role
Sidse Ehmsen et al.
Cell reports, 27(13), 3927-3938 (2019-06-27)
Tumor eradication may be greatly improved by targeting cancer stem cells (CSCs), as they exhibit resistance to conventional therapy. To gain insight into the unique biology of CSCs, we developed patient-derived xenograft tumors (PDXs) from ER- breast cancers from which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service