Skip to Content
Merck
All Photos(2)

Documents

H9882

Sigma-Aldrich

4-Hydroxybenzhydrazide

powder, ≥97%

Synonym(s):

4-Hydroxybenzoic hydrazide, 4-Hydroxybenzoylhydrazine, p-Hydroxybenzoic acid hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CONHNH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
473052
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Hydroxybenzhydrazide, ≥97%

biological source

synthetic

Quality Level

Assay

≥97%

form

powder

mp

264-266 °C (dec.) (lit.)

solubility

acetic acid: water (1:1): 25 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 360 nm; λem 425 nm (reacted with 5-hydroxymethyl-furaldehydezinc acetate)

SMILES string

NNC(=O)c1ccc(O)cc1

InChI

1S/C7H8N2O2/c8-9-7(11)5-1-3-6(10)4-2-5/h1-4,10H,8H2,(H,9,11)

InChI key

ZMZGIVVRBMFZSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Hydroxybenzhydrazide has been used:
  • for determining the concentration of maltose from starch digestion
  • for analysis of sugars from fructan containing food hydrolysate
  • in colorimetric assay for reducing sugar estimation from processed feedstuff

Biochem/physiol Actions

4-Hydroxybenzhydrazide (PAHBAH), is used for assaying α-amylase and glucoamylase activities. The assay based on PAHBAH measures non-specifically the free reducing sugar groups present in samples at 415nm colorimetrically.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Xie et al.
International journal of biological macromolecules, 126, 525-530 (2018-12-28)
Starch-containing food normally has complex components and structure, which control its digestion kinetic properties and nutritional value. However, interactions among multiple food components and their effects on starch digestion kinetics are rarely reported. Here, we evaluated the influence of tea
Extrusion induced low-order starch matrices: Enzymic hydrolysis and structure
Zhang B, et al.
Carbohydrate Polymers, 134(10), 485-496 (2015)
Dongling Qiao et al.
Carbohydrate polymers, 144, 271-281 (2016-04-17)
Combined analytical techniques were used to explore the effects of alkali treatment on the multi-scale structure and digestion behavior of starches with different amylose/amylopectin ratios. Alkali treatment disrupted the amorphous matrix, and partial lamellae and crystallites, which weakened starch molecular
Glucoamylase (exo-1, 4-alpha-d-glucan glucanohydrolase, EC 3.2. 1.3) is the major starch-degrading enzyme secreted by the phytopathogenic fungus Colletotrichum gloeosporioides
Krause DR, et al
Microbiology, 137(10), 2463-2468 (1991)
Efficacy of reducing sugar and phenol-sulfuric acid assays for analysis of soluble carbohydrates in feedstuffs
Hall, Mary Beth
Anim. Feed Sci. Technol., 185(1-2), 94-100 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service