Skip to Content
Merck
All Photos(3)

Key Documents

H9539

Sigma-Aldrich

Anti-HMGB1 (HMG1) (C-terminal) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Amphoterin, Anti-High mobility group box 1, Anti-High mobility group protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 5-10 μg using HEK-293T cell lysates.
indirect immunofluorescence: suitable
western blot: 0.5-1 μg/mL using 3T3 or PC12 cell lysates.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB1(3146)
mouse ... Hmgb1(15289)
rat ... Hmgb1(25459)

General description

The High Mobility Group (HMG1) is secreted by activated monocytes and macrophages. The gene is mapped to human chromosome 13q12.

Immunogen

synthetic peptide corresponding to amino acid 165-180 of human HMGB1, conjugated to KLH. The corresponding sequence is conserved in mouse and rat.

Application

Anti-HMGB1 (HMG1) (C-terminal) antibody produced in rabbit has been used in ELISA and immunofluorescence.

Biochem/physiol Actions

In the nucleus, HMGB1 (high-mobility group protein B1) works as a nucleosome stabilizer and transcriptional regulator. It also exhibits chaperone-like activity and suppresses aggregation of polyglutamine. It has a role in inflammation and autophagy. HMGB1 can interact with toll-like receptor (TLR) 2 and 4, interleukin-1β, chemokine (C-X-C motif) ligand 12 (CXCL12) and pathogen-associated molecular pattern (PAMP) molecules. It also works as a damage-associated molecular pattern molecule (DAMP).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, freeze in working aliquots.
Repeated freezing and thawing, or storage in “frostfree” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Other Notes

In order to obtain the best results using various techniques and preparations, we recommend determining the optimal working dilutions by titration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Qiu et al.
Journal of cellular and molecular medicine, 25(17), 8244-8260 (2021-08-20)
Septic cardiomyopathy is a common complication of sepsis with high morbidity and mortality, but lacks specific therapy. This study aimed to reveal the role of circTLK1 and its potential mechanisms in septic cardiomyopathy. The in vitro and in vivo models
MiR-142-3p functions as a potential tumor suppressor directly targeting HMGB1 in non-small-cell lung carcinoma.
Xiao P and Liu WL
International Journal of Clinical and Experimental Pathology (2015)
Liwei Weng et al.
Analytical chemistry, 90(12), 7552-7560 (2018-05-24)
High mobility group box 1 (HMGB1) is a non-histone chromosomal protein, which can be secreted through a variety of pathways and bind to pattern recognition receptors to release pro-inflammatory cytokines. Previous studies have suggested that HMGB1 is upregulated in numerous
Ross Pirnie et al.
Chemical research in toxicology, 35(10), 1893-1902 (2022-08-04)
The high mobility group box 1 (HMGB1), which is released during acute acetaminophen (APAP) overdose, is thought to mediate a subsequent immune response, particularly hepatic infiltration of macrophages. The redox behavior of HMGB1 and the proteoforms of HMGB1 present in
HMGB1 facilitates hypoxia-induced vWF upregulation through TLR2-MYD88-SP1 pathway
Bandana Singh, et al.
European Journal of Immunology (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service