Skip to Content
Merck
All Photos(1)

Key Documents

H7752

Sigma-Aldrich

Serotonin creatinine sulfate monohydrate

≥99% (TLC), powder, neurotransmitter

Synonym(s):

3-[2-Aminoethyl]-5-hydroxyindole creatinine sulfate complex, 5-HT, 5-Hydroxytryptamine creatinine sulfate complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19N5O2 · H2O4S · H2O
CAS Number:
Molecular Weight:
405.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Serotonin creatinine sulfate monohydrate, powder

form

powder

color

white to off-white

solubility

0.1 M HCl: soluble (Solutions should be freshly prepared.)
95% ethanol: very slightly soluble (Solutions should be freshly prepared.)
absolute ethanol: insoluble

storage temp.

2-8°C

SMILES string

O.OS(O)(=O)=O.CN1CC(=O)NC1=N.NCCc2c[nH]c3ccc(O)cc23

InChI

1S/C10H12N2O.C4H7N3O.H2O4S.H2O/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;1-7-2-3(8)6-4(7)5;1-5(2,3)4;/h1-2,5-6,12-13H,3-4,11H2;2H2,1H3,(H2,5,6,8);(H2,1,2,3,4);1H2

InChI key

BKCXVJIGPVULPX-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Application

Serotonin creatinine sulfate monohydrate has been used:
  • to study its effect on the feeding motions in Caenorhabditis elegans[1]
  • to study the chronotropic responses of serotonin (5-HT) in the zebrafish heart[2]
  • to study the effect of serotonin on isthmus peristalsis[3]

Biochem/physiol Actions

Neurotransmitter.
Serotonin creatinine sulfate stimulates neuronal production of catecholamine.[4]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuji Igarashi et al.
Journal of the American Chemical Society, 142(16), 7542-7554 (2020-04-15)
The rotation of an object cannot be fully tracked without understanding a set of three angles, namely, roll, pitch, and yaw. Tracking these angles as a three-degrees-of-freedom (3-DoF) rotation is a fundamental measurement, facilitating, for example, attitude control of a
Methods for measuring pharyngeal behaviors
WormBook: The Online Review of C. elegans Biology (2005)
Adolfo E Talpalar et al.
Nature, 500(7460), 85-88 (2013-07-03)
All forms of locomotion are repetitive motor activities that require coordinated bilateral activation of muscles. The executive elements of locomotor control are networks of spinal neurons that determine gait pattern through the sequential activation of motor-neuron pools on either side
Effect of serotonin on brown adipose tissue and on its sympathetic neurons
Steiner G and Evans S
The American Journal of Physiology, 231(1), 34-39 (1976)
Serotonin activates overall feeding by activating two separate neural pathways in Caenorhabditis elegans
Song B and Avery L
The Journal of Neuroscience, 32(6), 1920-1931 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service