Skip to Content
Merck
All Photos(1)

Key Documents

H7396

Sigma-Aldrich

(±)-2-Hydroxyoctanoic acid

Synonym(s):

2-Hydroxycaprylic acid, (±)-2-Hydroxycaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)COOH
CAS Number:
Molecular Weight:
160.21
Beilstein:
1760638
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCC(O)C(O)=O

InChI

1S/C8H16O3/c1-2-3-4-5-6-7(9)8(10)11/h7,9H,2-6H2,1H3,(H,10,11)

InChI key

JKRDADVRIYVCCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

(±)-2-Hydroxyoctanoic acid inhibits purified medium-chain acyl-CoA synthetase from bovine liver mitochondria (Ki, 500 uM) with hexanoic acid as a substrate .
(±)-2-Hydroxyoctanoic acid is a mixture of the D and L-2-hydroxyoctanoic acid enantiomers. (±)-2-Hydroxyoctanoic acid may be used in the development of medium-chain hydroxy-fatty acid enantiomer separation and detection systems. 2-Hydroxyoctanoic acid may be used as an inhibitor to study the kinteics of medium chain acyl-CoA synthetase(s).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane Grison et al.
Metabolomics : Official journal of the Metabolomic Society, 12(10), 154-154 (2016-10-13)
Data are sparse about the potential health risks of chronic low-dose contamination of humans by uranium (natural or anthropogenic) in drinking water. Previous studies report some molecular imbalances but no clinical signs due to uranium intake. In a proof-of-principle study
Yang Zhang et al.
Drug metabolism and pharmacokinetics, 34(3), 187-193 (2019-03-17)
Mandelic acid (MA) is generally used as a biomarker of the exposure of styrene, which is classified as a class of hazardous environmental pollutants, and also used as an important chiral intermediate in pharmaceutical industry. The previous studies have found
M Duran et al.
European journal of pediatrics, 150(3), 190-195 (1991-01-01)
Two siblings were found to be affected by long-chain 3-hydroxyacyl-CoA dehydrogenase deficiency, one of which died suddenly and unexpectedly on the 3rd day of life suffering from extreme hypoketotic hypoglycaemia. The younger sibling started to have feeding problems, lowered consciousness
Ying Li et al.
Plant physiology, 180(4), 1860-1876 (2019-05-30)
Mobilization of seed storage compounds, such as starch and oil, is required to provide energy and metabolic building blocks during seedling development. Over 50% of fatty acids in Arabidopsis (Arabidopsis thaliana) seed oil have a cis-double bond on an even-numbered
Worapan Pormsila et al.
Electrophoresis, 31(12), 2044-2048 (2010-05-25)
The enantiomers of the anions of five alpha-hydroxy acids, namely lactic acid, alpha-hydroxybutyric acid, 2-hydroxycaproic acid, 2-hydroxyoctanoic acid and 2-hydroxydecanoic acid, as well as the two alpha-amino acids aspartic acid and glutamic acid, were baseline separated and detected by CE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service