Skip to Content
Merck
All Photos(1)

Key Documents

H5413

Millipore

HIS-Select® Elution Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703
NACRES:
NA.56

Quality Level

General description

Elution buffer is a major solvent in affinity chromatography. Elution buffer is used to wash away unbound proteins at first and at a greater concentration it releases the desired protein from the ligand. It is important that the elution buffer works quickly without changing the function or activity of the desired protein.

Application

HIS-Select® Elution Buffer has been used in the elution of recombinant coagulase protein and tomoregulin 

Physical form

Contains 250 mM Imidazole, 0.3 M sodium chloride, 50 mM sodium phosphate, pH 8.0.

Legal Information

HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Seok Hong et al.
Journal of Alzheimer's disease : JAD, 48(3), 731-743 (2015-09-25)
Amyloid-β (Aβ) protein causes neurotoxicity and its abnormal aggregation into amyloid is a pathological hallmark of Alzheimer's disease (AD). Cellular proteins able to interact with Aβ or its precursor, AβPP (amyloid-β protein precursor), may regulate Aβ production and neurotoxicity. We
Affinity Chromatography
Scouten, William H
Chromatography (1981)
Alaa H Sewid et al.
Infection and immunity, 86(8) (2018-06-13)
Coagulase activation of prothrombin by staphylococcus induces the formation of fibrin deposition that facilitates the establishment of infection by Staphylococcus species. Coagulase activity is a key characteristic of Staphylococcus pseudintermedius; however, no coagulase gene or associated protein has been studied
Pär Comstedt et al.
PloS one, 9(11), e113294-e113294 (2014-11-20)
There is currently no Lyme borreliosis vaccine available for humans, although it has been shown that the disease can be prevented by immunization with an OspA-based vaccine (LYMErix). Outer surface protein A (OspA) is one of the dominant antigens expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service