Skip to Content
Merck
All Photos(3)

Key Documents

H1771

Sigma-Aldrich

3-Hydroxy-DL-kynurenine

Synonym(s):

α,2-Diamino-3-hydroxy-γ-oxobenzenebutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

1 M HCl: 49.00-51.00 mg/mL, clear to very slightly hazy

storage temp.

2-8°C

SMILES string

NC(CC(=O)c1cccc(O)c1N)C(O)=O

InChI

1S/C10H12N2O4/c11-6(10(15)16)4-8(14)5-2-1-3-7(13)9(5)12/h1-3,6,13H,4,11-12H2,(H,15,16)

InChI key

VCKPUUFAIGNJHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Hydroxy-DL-kynurenine is a tryptophan metabolite with a molecular weight corresponding to 224Da. It is a chromophore or hydrophilic yellow compound present in the lens of the eye. 3-Hydroxy-DL-kynurenine is synthesized from kynurenine by the action of enzyme kynurenine 3-monooxygenase (KMO). It is metabolized to 3-hydroxyanthranilic acid and xanthurenic acid in the presence of enzymes kynureninase and kynurenine aminotransferases, respectively.

Application

3-Hydroxy-DL-kynurenine has been used:
  • as a substrate for the recombinant human kynureninase assay
  • as a reference standard in tandem mass spectrometry (MS/MS) analysis
  • as a control for quantifying serum 3-Hydroxy-DL-kynurenine levels in diabetic retinopathy patients

Biochem/physiol Actions

3-Hydroxy-DL-kynurenine (3-HKYN) has antioxidant functionality and prevents lipid peroxidation in cerebral cortex. It may modulate synaptic neurotransmission. The levels of 3-HKYN is elevated in Huntington′s disease. However, its role in neurodegenerative diseases is still not understood well. Kynurenine metabolism is linked with glutamatergic neurotransmission and the level of 3-HKYN is linked to the pathophysiology of schizophrenia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd Bradley et al.
Journal of immunology (Baltimore, Md. : 1950), 197(12), 4663-4673 (2016-11-17)
The HIV-1 envelope protein (Env) has evolved to subvert the host immune system, hindering viral control by the host. The tryptophan metabolic enzyme kynureninase (KYNU) is mimicked by a portion of the HIV Env gp41 membrane proximal region (MPER) and
Mass spectrometric identification of urinary biomarkers of pulmonary tuberculosis
Isa F, et al.
EBioMedicine, 31(12), 157-165 (2018)
A M Myint et al.
Brain, behavior, and immunity, 25(8), 1576-1581 (2011-05-31)
The association between the pro-inflammatory state of schizophrenia and increased tryptophan degradation into kynurenine has been reported. However, the relationship between metabolites from subdivisions of the kynurenine pathway, kynurenic acid and 3-hydroxykynurenine, remains unknown. The present study tested the relationship
Yuri P Tsentalovich et al.
Investigative ophthalmology & visual science, 52(10), 7687-7696 (2011-08-30)
To compare the photochemical properties of UV filter molecules present in the human lens (kynurenine, KN; 3-hydroxykynurenine, 3OHKN; 3-hydroxykynurenine O-β-D-glucoside, 3OHKG; 4-(2-aminophenyl)-4-oxobutanoic acid, AHA; and glutathionyl-kynurenine, GSH-KN) with the use of the following parameters: excited singlet lifetime τ(S), fluorescence quantum
Sarah S Zaher et al.
Investigative ophthalmology & visual science, 52(5), 2640-2648 (2011-01-08)
IDO (indoleamine 2,3-dioxygenase) modulates the immune response by depletion of the essential amino acid tryptophan, and IDO overexpression has been shown to prolong corneal allograft survival. This study was conducted to examine the effect of kynurenines, the products of tryptophan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service