Skip to Content
Merck
All Photos(1)

Key Documents

H1136

Sigma-Aldrich

Hyaluronidase from Streptomyces hyalurolyticus

Synonym(s):

Hyaluronate Lyase from Streptomyces hyalurolyticus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Streptomyces sp. (S. hyalurolyticus)

Quality Level

conjugate

conjugate (Glucosaminoglycan)

form

film (Clear)

packaging

ampule of 300 ×  units

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hyaluronidase is widely present in nature. In humans, it is present in a number of organs and body fluids. Hyaluronidase is found in the secretions of virus, bacteriophage, fungi and bacteria. It is also present in nematode and leeches secretions. Snakes and scorpions produce hyaluronidase in their venom. Hyaluronidase is also secreted by bees, spiders, wasps, hornets, caterpillars, fishes and lizards.

Application

Hyaluronidase from Streptomyces hyalurolyticus has been used:
  • in the preparation of DMEM/F-12 (Dulbecco′s modified eagle medium/nutrient mixture F-12) media for the isolation and purification of single cells from dissociated tumour tissue
  • as a component of digestion solution for the derivation of germline stem cells from testicular tissue

Biochem/physiol Actions

Hyaluronate lyase cleaves hyaluronic acid at the β-D-GlcNAc-(1→4)-β-D-GlcA bond, yielding 3-(4-deoxy-β-D-gluc-4-enuronosyl)-N-acetyl-D-glucosamine tetra- and hexasaccharides. Unlike other hyaluronidases, this enzyme is specific for hyaluronic acid and is inactive with chondroitin and chondroitin sulfate.
Hyaluronidase is an endoglycosidase and performs its function along with exoglycosidases, like β-glucuronidase and β-N-acetyl glucosaminidase. Hyaluronidase regulates the hyaluronic acid turnover.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of hyaluronidase, a major allergen of bee venom
Markovic H, et al.
Structure, 8(10), 1025-1035 (2000)
Connor C Gains et al.
Frontiers in bioengineering and biotechnology, 8, 700-700 (2020-08-09)
The Achilles tendon (AT) is comprised of three distinct sub-tendons bound together by the inter-subtendon matrix (ISTM). The interactions between sub-tendons will have important implications for AT function. The aim of this study was to investigate the extent to which
Wen Min Lau et al.
Cancer research, 74(9), 2630-2641 (2014-03-13)
The surface marker CD44 has been identified as one of several markers associated with cancer stem cells (CSC) in solid tumors, but its ubiquitous expression in many cell types, including hematopoietic cells, has hindered its use in targeting CSCs. In
Tímea Váradi et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 60(8), 567-575 (2012-05-09)
Although trastuzumab is an efficient drug, primary and acquired resistance is a challenging problem. The authors have previously shown in mouse xenograft experiments that masking ErbB2 by hyaluronan leads to diminished binding of the antibody and consequent resistance. In the
Conversion of adult mouse unipotent germline stem cells into pluripotent stem cells
Ko K, et al.
Nature Protocols, 5(5), 921-921 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service