Skip to Content
Merck
All Photos(1)

Documents

H0784

Sigma-Aldrich

Hybri-slips

L × W × thickness 60 mm × 24 mm × 0.25 mm

Synonym(s):

hybridization cover, polycarbonate hybridization cover

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

polycarbonate

packaging

pack of 100 ea

manufacturer/tradename

Grace Bio-Labs 716024

L × W × thickness

60 mm × 24 mm × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Constant probe concentration - Unlike glass cover slips, Hybri-slips do not adsorb probes.
  • Ready-to-use - Hybri-slips do not require pre-treatment prior to use.
  • Avoid RNA degradation - Hybri-slips are supplied RNAse-free and are protected from contamination by clean-release liners.
  • Durable - Hybri-slips remain flat even at elevated temperatures.

Application

Hybri-slips have been used on slides in microarray hybridization. They have also been used for in situ hybridization.
Hybri-slips are hydrophobic cover slips for in situ hybridization, in situ PCR, and hybridization to genomic arrays on glass slides.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Expression patterns of Passiflora edulis APETALA1/FRUITFULL homologues shed light onto tendril and corona identities.
Scorza LC, et al.
EvoDevo, 8, doi: 10-doi: 10 (2017)
The tagged microarray marker (TAM) method for high-throughput detection of single nucleotide and indel polymorphisms.
Jing R, et al.
Nature Protocols, 2, 168-177 (2007)
Rachele Macirella et al.
International journal of molecular sciences, 17(3), 361-361 (2016-03-16)
Mercury (Hg) is a highly hazardous pollutant widely used in industrial, pharmaceutical and agricultural fields. Mercury is found in the environment in several forms, elemental, inorganic (iHg) and organic, all of which are toxic. Considering that the liver is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service