Skip to Content
Merck
All Photos(1)

Key Documents

GW22461F

Sigma-Aldrich

Anti-Interleukin-2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-22461F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL2(3558)

General description

IL2 (interleukin 2) is a pro-inflammatory cytokine, which is predominantly synthesized by activated T-cells and dendritic cells (DCs). T-cells also act as effector of IL-2 as they contain the interleukin 2 receptor (IL2R). This gene is localized to human chromosome 4q27, and codes for a glycoprotein with a molecular weight of 14-17kDa.

Immunogen

Immunogen Sequence: GI # 28178861, sequence 1-153
Recombinant interleukin 2

Application

Anti-Interleukin-2 antibody produced in chicken is suitable for the production of anti-IL2 surface-conjugated microtoroids. It is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Interleukin 2 (IL-2) is an interleukin and is a cytokine signaling molecule in the immune system. It plays an important role in signal transduction for biological functions of IL-2. Upregulation of IL-2 combined with activation of IL-2-dependent signaling pathways may contribute to the pathogenesis of Focal cortical dysplasia (FCD). Aerosol interleukin-2 (IL-2) increases the number of intravenously injected human natural killer (NK) cells in the lungs by stimulating local NK cell proliferation. The effect on NK cell proliferation is organ specific, which makes it ideal for the specific targeting of lung metastasis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hunter A Martinez et al.
Nature communications, 15(1), 1564-1564 (2024-02-21)
Although FOXP3+ regulatory T cells (Treg) depend on IL-2 produced by other cells for their survival and function, the levels of IL-2 in inflamed tissue are low, making it unclear how Treg access this critical resource. Here, we show that
Wei Guo et al.
Journal of neuropathology and experimental neurology, 73(3), 206-222 (2014-02-04)
Focal cortical dysplasia (FCD) is a well-known cause of medically intractable epilepsy. To understand the potential role of the inflammatory cytokine interleukin 2 (IL-2) in the pathogenesis of FCD, we investigated the expression patterns of IL-2 and its receptors (IL-2Rs)
Marcel Joly et al.
Journal of theoretical biology, 335, 57-78 (2013-06-29)
In this paper, we sought to identify the CD4(+) T-cell dynamics in the course of HIV infection in response to continuous and intermittent intravenous courses of interleukin-2 (IL-2), the principal cytokine responsible for progression of CD4(+) T-lymphocytes from the G1
Marta Fichna et al.
Molecular biology reports, 40(12), 6957-6963 (2013-10-25)
Polymorphic variants at the interleukin-2 (IL2) locus affect the risk of several autoimmune disorders. Our aim was to evaluate the association of the four IL2 polymorphisms (rs6822844, rs6534349, rs2069762 and rs3136534) with type 1 diabetes (T1D) in the Polish population
Jipseol Jeong et al.
Veterinary immunology and immunopathology, 162(3-4), 154-161 (2014-12-03)
Interleukin (IL)-2 and IL-15 receptor β (IL-2/15Rβ, CD122) play important roles in signal transduction for biological functions of IL-2 and IL-15. We found that ducks possess three different IL-2/15Rβ transcripts, a conventional form (duIL-2/15Rβ) and two variants. Comparisons between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service