Skip to Content
Merck
All Photos(1)

Key Documents

GERPN226

cGMP, Biotrak EIA System

Cytiva RPN226

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116126
NACRES:
NA.32
Pricing and availability is not currently available.

manufacturer/tradename

Cytiva RPN226

packaging

pkg of 96 wells

storage temp.

2-8°C

Related Categories

General description

EIAs and ELISAs are convenient, high-sensitivity, peroxidase-based technologies that enable the amount of protein present in a sample to be measured in a single day. Novel lysis reagents have been developed (Lysis Reagents 1 and 2) to facilitate simple and rapid extraction of cellular molecules.

The Biotrak cGMP competitive enzymeimmunoassay system is specifically designed for research purposes. The kit includes protocols using novel lysis reagents in order to facilitate simple and rapid extraction of cGMP from cell cultures. These components avoid the requirement for traditional, time-consuming extraction procedures and obviate the need for removal of extraction reagents prior to measurement. It combines the use of a peroxidase-labeled cGMP conjugate, a specific antiserum which can be immobilized on to precoated microplates, and a one-pot stabilized substrate solution.

Lysis reagent 1 hydrolyzes cell membranes to release intra-cellular cAMP. Lysis Reagent 2 sequesters the key component in Lysis Reagent 1 and ensures cAMP is free for subsequent analysis. The detergent/sequestrant complex does not interfere with antigen: antibody binding. Lysis Reagent 1 is simply added to cultured cells, followed by a 5 to 10 min incubation before assay. The antiserum is reconstituted with Lysis Reagent 2. The assay is based on competition between unlabeled cAMP and a fixed quantity of peroxidase-labeled cAMP, for a limited number of binding sites on a cAMP specific antibody.

Application

cGMP, Biotrak EIA System has been used for measuring total cellular cyclic GMP (cGMP) or cyclic AMP (cAMP) levels.[1][2]

Features and Benefits

  • Elimination of inconvenient, time-consuming extraction procedures.
  • Flexible method-choice of assay protocols.
  • Dual range 2–512 fmol/well (acetylation protocol), dual range 50–12 800 fmol/well (non-acetylation protocol).
  • Rapid assay protocol.
  • Nonradioactive.
  • Specific for cGMP
  • Precise and accurate measurement.
  • Ready-to-use substrate.
  • Colour coded reagents

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak is a trademark of Cytiva

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

YC-1 potentiates the nitric oxide/cyclic GMP pathway in corpus cavernosum and facilitates penile erection in rats.
Gin CH, et al.
European Journal of Pharmacology, 458(1-2), 183-189 (2003)
Ca2+ -permeable AMPA (-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic Acid) Receptors and Dopamine D1 Receptors Regulate GluA1 Trafficking in Striatal Neurons
David S T and Edward B Z
The Journal of Biological Chemistry, 288(9) (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service