Skip to Content
Merck
All Photos(1)

Key Documents

GENA931-1ML

Mouse IgG HRP Linked Whole Ab

Cytiva NA931-1ML

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.76

feature

wetted part: no

packaging

pkg of 1 mL

manufacturer/tradename

Cytiva NA931-1ML

color

Brown (light)

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

The HRP Linked whole antibodies are prepared by hyper-immunizing an animal with purified immunoglobulin fractions from normal human serum to produce high-affinity antibodies.

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.

Application

Mouse IgG HRP Linked Whole Ab has been used in immunoblotting

Features and Benefits

  • Highly species-specific.
  • Optimized for use with our range of ECL Detection Reagents.
  • Recommended dilutions to minimize background.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reliable set of reference genes to normalize oxygen-dependent cytoglobin gene expression levels in melanoma
De Backer J, Maric D
Scientific Reports, 11(1), 10879-10879 (2021)
Joey De Backer et al.
Scientific reports, 11(1), 10879-10879 (2021-05-27)
Cytoglobin (CYGB) is a ubiquitously expressed protein with a protective role against oxidative stress, fibrosis and tumor growth, shown to be transcriptionally regulated under hypoxic conditions. Hypoxia-inducible CYGB expression is observed in several cancer cell lines and particularly in various
Valéna Karaghiannis et al.
Haematologica (2023-01-27)
Gain-of-function mutations in the EPAS1/HIF2A gene have been identified in patients with hereditary erythrocytosis that can be associated with the development of paraganglioma, pheochromocytoma and somatostatinoma. In the present study, we describe a unique european collection of 41 patients and
Anna Keppner et al.
International journal of molecular sciences, 23(13) (2022-07-10)
The kidney is strongly dependent on a continuous oxygen supply, and is conversely highly sensitive to hypoxia. Controlled oxygen gradients are essential for renal control of solutes and urine-concentrating mechanisms, which also depend on various hormones including aldosterone. The cortical
Elvira Nikalayevich et al.
Developmental cell, 59(7), 841-852 (2024-02-23)
The cortex controls cell shape. In mouse oocytes, the cortex thickens in an Arp2/3-complex-dependent manner, ensuring chromosome positioning and segregation. Surprisingly, we identify that mouse oocytes lacking the Arp2/3 complex undergo cortical actin remodeling upon division, followed by cortical contractions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service