Skip to Content
Merck
All Photos(1)

Key Documents

GE29-0181-82

HiPrep Q High Performance 16/10

Cytiva 29-0181-82, pack of 1 column

Synonym(s):

HiPrep Q Column, High Performance Q Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pack of 1 column

manufacturer/tradename

Cytiva 29-0181-82

parameter

1-5 ml/min flow rate
44 psi

bed size

16 mm × 100 mm

column I.D.

16 mm

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning

1-14

working range

2-12

suitability

suitable for bioprocess medium

General description

HiPrep Q HP is a 20 mL strong anion exchange chromatography column for high resolution small-scale protein purification. The Q functional group is a quaternary amino group coupled to the matrix via chemically stable ether linkages. Q Sepharose High Performance is a strong anion exchanger charged with high loading capacity over broad pH ranges.

Features and Benefits

  • Packed with Q Sepharose High Performance strong quarternary ammonium anion exchange resin.
  • Small (34 µm) bead size delivers high-performance, high-resolution purifications.
  • 20 mL HiPrep column format compatible with single-pump configurations and chromatography systems such as ÄKTA systems.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiPrep is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service