GE17-5468-10
Capto™ SP ImpRes
Cytiva 17-5468-10, pack of 25 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
ligand
sulfonate group
description
Ion Exchanger Type (value)
packaging
pack of 25 mL
manufacturer/tradename
Cytiva 17-5468-10
matrix
high-flow agarose
avg. part. size
36-44 μm (d50v)
cleaning in place
3-14
working range
4-12
capacity
>70 mg binding capacity(> 70 mg lysozyme, > 95 mg BSA)
suitability
suitable for bioprocess medium
Related Categories
General description
Capto™ SP ImpRes is a strong cation exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.
Application
Capto™ SP ImpRes is a strong cation exchanger for the high-throughput intermediate purification and polishing steps of a wide range of biomolecules. This chromatography media is part of an expanded high-resolution platform based on the high-fLow Agarose Capto™ product line.
By combining the high-fLow characteristics of Capto™ media with a small particle size, Capto™ SP ImpRes delivers excellent pressure-fLow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design.
Capto™ SP ImpRes is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical Proteins. Based on the well-established Capto™ base matrix and with traditional ionic group ligands, it provides many opportunities for improved productivity and straightforward process development.
Capto™ SP ImpRes is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto™ SP ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.
By combining the high-fLow characteristics of Capto™ media with a small particle size, Capto™ SP ImpRes delivers excellent pressure-fLow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design.
Capto™ SP ImpRes is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical Proteins. Based on the well-established Capto™ base matrix and with traditional ionic group ligands, it provides many opportunities for improved productivity and straightforward process development.
Capto™ SP ImpRes is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto™ SP ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- High-resolution intermediate purification and polishing media based on the well-established Capto™ platform with traditional ligands.
- Flexible process design due to large operational window of flow rates and bed heights.
- High throughput purifications easy to optimize and scale-up.
- Higher manufacturing productivity enables improved process economy.
- BioProcess media developed and supported for large-scale chromatography.
- Prepacked HiTrap® columns for easy screening and small-scale purification.
- Security of supply and comprehensive regulatory support
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service