GE17-5316-10
Capto™ Q
Cytiva 17-5316-10, pack of 25 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47101511
NACRES:
NA.56
Recommended Products
ligand
quaternary amine
description
Ion Exchanger Type (value)
packaging
pack of 25 mL
manufacturer/tradename
Cytiva 17-5316-10
matrix
highly cross-linked agarose with dextran surface extender
average diameter
90 μm (d50v)
cleaning in place
2-14
working range
2-12
suitability
suitable for bioprocess medium
General description
Capto™ Q is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification.
Application
Capto™ Q is designed for capture and intermediate purification of Proteins from large feed volumes. It is composed of a rigid, high-fLow Agarose matrix modified with Dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto™ Q raises the productivity in industrial downstream processes. Capto™ Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
Capto™ Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto™ Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Capto™ Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto™ Q meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Strong anion exchanger for high-productivity capture and intermediate purification when high volume throughput is essential.
- High dynamic binding capacity at high flow raises productivity
- High-volume throughput cuts process times
- Cost-effective processing with smaller unit operations.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Capto is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service