GE17-5269-02
MabSelect™ Xtra
Cytiva 17-5269-02, pack of 200 mL
About This Item
Recommended Products
ligand
recombinant protein A (E. coli)
packaging
pack of 200 mL
manufacturer/tradename
Cytiva 17-5269-02
storage condition
(20% Ehtanol)
matrix
highly cross-linked agarose
avg. part. size
75 (d₅₀v is the median particle size of the cumulative volume distribution)
cleaning in place
2-12
working range
3-10
capacity
≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)
suitability
suitable for bioprocess medium
storage temp.
2-8°C
General description
Application
The medium provides a lower overall production cost due to the possibility of processing concentrated feedstocks in fewer batches.
MabSelect™ Xtra is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect™ Xtra meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Higher dynamic binding capacity than currently available Protein A chromatography media.
- Improved process economics through reduced raw materials costs and/or reduced number of cycles
- High purity capture due to high ligand selectivity and matrix hydrophilicity
- Slightly lower pressure/fLow properties compared to MabSelect™ SuRe™ and MabSelect
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service