GE11-0034-94
HiTrap® MabSelect SuRe™
Cytiva 11-0034-94, pack of 1 × 5 mL
Synonym(s):
HiTrap MabSelect SuRe, MabSelect SuRe Column
About This Item
Recommended Products
ligand
alkali-stabilized protein A-derived (E. coli)
packaging
pack of 1 × 5 mL
manufacturer/tradename
Cytiva 11-0034-94
storage condition
(20% Ehtanol)
(20% Ethanol)
parameter
<20 mL/min flow rate
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
highly cross-linked agarose
avg. part. size
85 μm (d₅₀v is the median particle size of the cumulative volume distribution)
cleaning
2-13.7
working range
3-12
capacity
≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)
suitability
suitable for bioprocess medium
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
HiTrap® MabSelect™ columns are prepacked with MabSelect™ a bioprocess medium for capuring of Mabs from large sample volumes.
Features and Benefits
- Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
- Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage.
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity.
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
- Prepacked HiTrap® columns for process development, screening of purification conditions, and small-scale purification of Mabs.
Analysis Note
Other Notes
Legal Information
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service