GE10600029
Amersham™ Hybond® P Western blotting membranes, PVDF
pore size 0.45 μm, roll W × L 150 mm × 4 m, pkg of 1 ea
Synonym(s):
PVDF Blotting Membrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 10600029
roll W × L
150 mm × 4 m
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
General description
PVDF membranes have higher mechanical strength than NC membranes and, thus, are more suitable for Western blotting applications where stripping and reprobing is needed. In addition, PVDF membranes exhibit higher protein binding capacity, however, with a tendency for a higher background, compared with NC membranes.
Features and Benefits
- Offers high physical strength.
- For use with standard colorimetric and chemiluminescent detection methods for proteins of > Mr 20 000.
- Offers protein binding capacity over 200 μg/cm2.
- Even after rigorous washing processes, high protein retention is achieved.
- Sample loss is reduced by high protein capture during transfer.
- For laboratory use, it is available in a variety of precut sizes.
- Suitable for stripping and reprobing
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service