Skip to Content
Merck
All Photos(1)

Documents

GE10600016

Amersham Protran® Supported Western blotting membranes, nitrocellulose

pore size 0.45 μm, roll W × L 300 mm × 4 m, pkg of 1 ea

Synonym(s):

Supported Nitrocellulose Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600016

roll W × L

300 mm × 4 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Amersham Protran® Supported Western blotting membranes, nitrocellulose, a robust protein blotting membrane, is made of reinforced nitrocellulose for multiple reprobings.

Application

Amersham Protran® Supported Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • High binding capacity.
  • Versatile, robust and excellent signal-to-noise ratios.
  • Optimal for applications when stripping and reprobing is required.
  • Suitable for proteins of a wide range of molecular weights.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service