GBL664508
Grace Bio-Labs Press-To-Seal silicone isolator, No PSA
wells, 24 (Round, 2mm diameter, 0.5mm depth)
Synonym(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
material
red silicone
sterility
non-sterile
feature
wells 24 (Round, 2mm diameter, 0.5mm depth)
packaging
pack of 25 ea
manufacturer/tradename
Grace Bio-Labs 664508
external size
25 mm × 54 mm
wells
24
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Application
Live-cell imaging
Microscoscopy
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service