Skip to Content
Merck
All Photos(1)

Key Documents

G8147

Sigma-Aldrich

Glucagon-Like Peptide I Amide Fragment 7-36 human

≥97% (HPLC), powder

Synonym(s):

Preproglucagon 78-107 Amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C149H226N40O45
CAS Number:
Molecular Weight:
3297.63
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)Cc4c[nH]cn4)[C@@H](C)O)[C@@H](C)O)C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(N)=O

InChI

1S/C149H226N40O45/c1-17-76(10)119(146(232)167-80(14)126(212)175-104(60-86-63-159-91-36-25-24-35-89(86)91)136(222)177-100(56-73(4)5)137(223)186-117(74(6)7)144(230)174-93(37-26-28-52-150)128(214)160-65-110(197)168-92(122(154)208)39-30-54-158-149(155)156)188-138(224)102(57-83-31-20-18-21-32-83)178-133(219)98(47-51-115(204)205)173-132(218)94(38-27-29-53-151)170-124(210)78(12)164-123(209)77(11)166-131(217)97(44-48-109(153)196)169-111(198)66-161-130(216)96(46-50-114(202)203)172-134(220)99(55-72(2)3)176-135(221)101(59-85-40-42-88(195)43-41-85)179-141(227)106(68-190)182-143(229)108(70-192)183-145(231)118(75(8)9)187-140(226)105(62-116(206)207)180-142(228)107(69-191)184-148(234)121(82(16)194)189-139(225)103(58-84-33-22-19-23-34-84)181-147(233)120(81(15)193)185-112(199)67-162-129(215)95(45-49-113(200)201)171-125(211)79(13)165-127(213)90(152)61-87-64-157-71-163-87/h18-25,31-36,40-43,63-64,71-82,90,92-108,117-121,159,190-195H,17,26-30,37-39,44-62,65-70,150-152H2,1-16H3,(H2,153,196)(H2,154,208)(H,157,163)(H,160,214)(H,161,216)(H,162,215)(H,164,209)(H,165,213)(H,166,217)(H,167,232)(H,168,197)(H,169,198)(H,170,210)(H,171,211)(H,172,220)(H,173,218)(H,174,230)(H,175,212)(H,176,221)(H,177,222)(H,178,219)(H,179,227)(H,180,228)(H,181,233)(H,182,229)(H,183,231)(H,184,234)(H,185,199)(H,186,223)(H,187,226)(H,188,224)(H,189,225)(H,200,201)(H,202,203)(H,204,205)(H,206,207)(H4,155,156,158)/t76-,77-,78-,79-,80-,81+,82+,90-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,117-,118-,119-,120-,121-/m0/s1

InChI key

DTHNMHAUYICORS-KTKZVXAJSA-N

Gene Information

human ... GCG(2641)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-NH2

General description

Glucagon-like peptide 1 (GLP-1) peptide is obtained from proglucagon and is metabolically inactive. However, it is processed to two smaller amide forms, namely GLP-1 (7-37) amide and GLP-1 (7-36) amide. It is a natural gastrointestinal peptide.[1]

Application

Glucagon-Like Peptide (GLP) I Amide Fragment 7-36 human has been used as a GLP-1 receptor agonist:
  • in lysate transfer cyclic adenosine monophosphate (cAMP) assay to test its effect on adenylyl cyclase activity[2]
  • to pre-treat the human umbilical vein endothelial cells (HUVECs) to monitor its effect on hyperpermeability in endothelial cell (EC) monolayers and changes with the actin cytoskeleton[3]
  • to test its inhibitory effect on vascular endothelial growth factor-A (VEGFA) based vasodilation in rat mesenteric arteries[4]

Glucagon-Like Peptide I Amide Fragment 7-36 human has been used as a supplement in DMEM (Dulbeccos′) medium for the culture of 3T3-L1 adipocytes to study the relation between adipose tissue GLP-1 Receptor and obesity and insulin resistance.[5]

Biochem/physiol Actions

GLP-1 (glucagon-like peptide 1) (7-36) amide is a potent insulinotropic peptide, which initiates the glucose-induced insulin release after meals or oral glucose intake. It has an anti-diabetogenic effect and thus, might have use in the treatment of non-insulin dependent diabetes mellitus.[1]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W G Ding et al.
Experimental physiology, 86(3), 331-339 (2001-06-29)
The present investigation was designed to examine whether calmodulin is involved in the inhibition of the ATP-sensitive K+ (K(ATP)) channel by glucagon-like peptide 1(7-36) amide (GLP-1) in mouse pancreatic beta-cells. Membrane potential, single channel and whole-cell currents through the K(ATP)
Nizar I Mourad et al.
Endocrinology, 153(10), 4644-4654 (2012-09-06)
Insulin secretion is triggered by an increase in the cytosolic calcium concentration ([Ca(2+)](c)) in β-cells. Ca(2+)-induced exocytosis of insulin granules can be augmented by metabolic amplification (unknown signals generated through glucose metabolism) or neurohormonal amplification (in particular cAMP mediated). Functional
M Gutniak et al.
The New England journal of medicine, 326(20), 1316-1322 (1992-05-14)
Glucagon-like peptide-1 (7-36) amide (glucagon-like insulinotropic peptide, or GLIP) is a gastrointestinal peptide that potentiates the release of insulin in physiologic concentrations. Its effects in patients with diabetes mellitus are not known. We compared the effect of an infusion of
Song-Tao Tang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 113, 108667-108667 (2019-03-11)
Glucagon-like peptide-1 (GLP-1) showed protective effects on endothelium-dependent dilatation. Since endothelial barrier dysfunction also plays a pivotal role in atherosclerosis, this study was designed to investigate the effects of GLP-1 on endothelial barrier function in diabetic aortic endothelium and explore
Haokao Gao et al.
Molecular pharmaceutics, 8(5), 1775-1782 (2011-08-02)
Derived from endocrine pancreatic beta cells, insulinomas express glucagon-like peptide-1 (GLP-1) receptor with high density and incidence. In this study, we labeled a novel GLP-1 analogue, EM3106B, with (18)F and performed PET imaging to visualize insulinoma tumors in an animal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service