Skip to Content
Merck
All Photos(3)

Key Documents

G7882

Sigma-Aldrich

L-Glutamic Dehydrogenase from bovine liver

Type III, lyophilized powder, ≥20 units/mg protein

Synonym(s):

L-GLDH, L-Glutamate:NAD[P]+ Oxidoreductase (deaminating), Glutamate Dehydrogenase from bovine liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine liver

Quality Level

type

Type III

form

lyophilized powder

specific activity

≥20 units/mg protein

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-Glutamic Dehydrogenase was used to catalyzes the conversion of isocitrate into a-ketoglutarate and carbon dioxide.[1]

Biochem/physiol Actions

Mammalian forms of this enzyme, including this bovine form, can use either NADP(H) or NAD(H) as coenzymes. L-glutamic dehydrogenase plays a unique role in mammalian metabolism. The reverse reaction catalyzed by this enzyme is the only pathway by which ammonia can become bound to the α-carbon atom of an α-carboxylic acid and thus, is the only source of de novo amino acid synthesis in mammalian species.

The bovine enzyme is characterized by three sets of properties:
  • It has a reversible concentration-dependent association, producing higher molecular weight forms.
  • Forms tight enzyme-reduced coenzyme-substrate ternary complexes whose rates of dissociation modulate the steady-state reaction rates.
  • Exhibits a wide variety of effects from the binding of any of a number of nucleotide modifiers.

L-glutamic dehydrogenase catalyzes the conversion of glutamate to α-ketoglutarate.

Packaging

Package size based on protein content

Unit Definition

One unit will reduce 1.0 μmole of α-ketoglutarate to L-glutamate per min at pH 7.3 at 25 °C, in the presence of ammonium ions.

Physical form

Contains citrate and potassium phoshate buffer salts.

Analysis Note

Protein determined by biuret

substrate

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roy M Daniel et al.
The Biochemical journal, 425(2), 353-360 (2009-10-24)
Experimental data show that the effect of temperature on enzymes cannot be adequately explained in terms of a two-state model based on increases in activity and denaturation. The Equilibrium Model provides a quantitative explanation of enzyme thermal behaviour under reaction
Laszlo Tretter et al.
Journal of neurochemistry, 83(4), 855-862 (2002-11-08)
Previously we have reported that oxidative stress induced by hydrogen peroxide exacerbates the effect of an Na+ load in isolated nerve terminals, with a consequence of an ATP depletion, [Ca2+]i and [Na+]i deregulation, and collapse of mitochondrial membrane potential. In
Decreased carbohydrate metabolism enzyme activities in the glaucomatous trabecular meshwork
Junk AK, Goel M, Mundorf T, Rockwood EJ, Bhattacharya SK
Molecular Vision, 10, 1286-1291 (2010)
Aidan P France et al.
Analytical chemistry, 92(6), 4340-4348 (2020-02-14)
Careful transfer of ions into the gas-phase permits the measurement of protein structures, with ion mobility-mass spectrometry, which provides shape and stoichiometry information. Collision cross sections (CCS) can be obtained from measurements made of the ions mobility through a given
S M Kuo et al.
Biology of the neonate, 43(1-2), 23-32 (1983-01-01)
The tissue distribution, the subcellular distribution in liver, and the developmental patterns of cysteine:alpha-ketoglutarate aminotransferase (CAT) and 3-mercaptopyruvate sulfurtransferase (MPST) activities were determined in rats of the Sprague-Dawley strain. CAT activity was highest in heart and liver, whereas MPST activity

Questions

  1. L-グルタミン酸デヒドロゲナーゼ ウシ肝臓由来(製品番号G7882)のKm値(Properties)や至適pH(Specificity)の情報を提供いただけませんでしょうか。

    1 answer
    1. The Km values for this product have not been determined. Please see the link below to review a reference that may be helpful:
      https://www.jbc.org/article/S0021-9258(19)83816-4/pdf

      The activity of this enzyme is determined at pH 7.3 at 25 °C. Please see the link below to review the product assay protocol:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/137/309/g7882enz.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service