Skip to Content
Merck
All Photos(2)

Documents

G7627

Sigma-Aldrich

Guanosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt

≥90%

Synonym(s):

MDCC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N6O8P · C17H31N3O
CAS Number:
Molecular Weight:
725.77
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥90%

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

C1CCC(CC1)N\C(=N\C2CCCCC2)N3CCOCC3.NC4=NC(=O)c5ncn([C@@H]6O[C@H](COP(O)(=O)N7CCOCC7)[C@@H](O)[C@H]6O)c5N4

InChI

1S/C17H31N3O.C14H21N6O8P/c1-3-7-15(8-4-1)18-17(20-11-13-21-14-12-20)19-16-9-5-2-6-10-16;15-14-17-11-8(12(23)18-14)16-6-20(11)13-10(22)9(21)7(28-13)5-27-29(24,25)19-1-3-26-4-2-19/h15-16H,1-14H2,(H,18,19);6-7,9-10,13,21-22H,1-5H2,(H,24,25)(H3,15,17,18,23)/t;7-,9-,10-,13-/m.1/s1

InChI key

CFNWUGDJWKRMBL-VAMAKUSHSA-N

Application

Guanosine 5′-monophosphomorpholidate may be used to synthesize guanosine diphosphate sugars such as GDP-fucose, GDP-mannose, UDP-galactose, and other more complex sugars nucleotides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vi N Nguyen et al.
Proteins, 84(12), 1810-1822 (2016-09-13)
The nearly 50,000 known Nudix proteins have a diverse array of functions, of which the most extensively studied is the catalyzed hydrolysis of aberrant nucleotide triphosphates. The functions of 171 Nudix proteins have been characterized to some degree, although physiological
Anting Xu et al.
Analytical biochemistry, 437(2), 178-184 (2013-03-14)
The common substrate structure for the functionally diverse Nudix protein superfamily is nucleotide-diphosphate-X, where X is a large variety of leaving groups. The substrate specificity is known for less than 1% of the 29,400 known members. Most activities result in
Tamami Koyama et al.
Journal of gerontological nursing, 42(10), 21-29 (2016-09-27)
HOW TO OBTAIN CONTACT HOURS BY READING THIS ARTICLE INSTRUCTIONS 1.3 contact hours will be awarded by Villanova University College of Nursing upon successful completion of this activity. A contact hour is a unit of measurement that denotes 60 minutes
Manika Indrajit Singh et al.
Biochimica et biophysica acta, 1861(1 Pt A), 3300-3310 (2016-08-25)
DNA polymerase processivity factors are ubiquitously present in all living organisms. Notwithstanding their high significance, the molecular details of clamps pertaining to the factors contributing to their stability are presently lacking. The bacteriophage T4 sliding clamp gp45 forms a homotrimer
Maria Mills et al.
Nucleic acids research, 45(20), 11878-11890 (2017-10-24)
The single-stranded DNA binding protein (SSB) of Escherichia coli plays essential roles in maintaining genome integrity by sequestering ssDNA and mediating DNA processing pathways through interactions with DNA-processing enzymes. Despite its DNA-sequestering properties, SSB stimulates the DNA processing activities of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service