Skip to Content
Merck
All Photos(2)

Key Documents

G6923

Sigma-Aldrich

Genapol® X-100

Synonym(s):

Isotridecanol ethoxylate, Isotridecyl alcohol polyglycol ether (10 EO), PEG isotridecyl ether, Polyethylene glycol monoisotridecyl ether, Polyoxyethylene (10) Isotridecyl Ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

Quality Level

mol wt

micellar average mol wt 56000

aggregation number

88

CMC

0.15 mM (20-25°C)
19.5

transition temp

cloud point 82 °C

HLB

13 - 14

InChI

1S/C30H62O10/c1-2-3-4-5-6-7-8-9-10-11-13-32-15-17-34-19-21-36-23-25-38-27-29-40-30-28-39-26-24-37-22-20-35-18-16-33-14-12-31/h31H,2-30H2,1H3

InChI key

ONJQDTZCDSESIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Genapol X-100 has been used in a study to assess how detergents affect inhibitor potencies against cyclo-oxygenase isoforms. It has also been used in a study to investigate the solubilisation of the membrane-bound D-alanyl-D-alanine carboxypeptidase of Bacillus coagulans.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Ouellet et al.
The Biochemical journal, 377(Pt 3), 675-684 (2003-09-27)
The sensitivity of Coxs (cyclo-oxygenases) to inhibition is known to be highly dependent on assay conditions. In the present study, the inhibitor sensitivities of purified Cox-1 and -2 were determined in a colorimetric assay using the reducing agent N, N
C Padrín-Sanz et al.
Journal of chromatography. A, 1078(1-2), 13-21 (2005-07-13)
A new methodology based on the microwave assisted micellar extraction (MAME) technique has been optimised, using soil samples, to extract and determine a mixture of the eight organophosphorous pesticides mainly used in agriculture. The pesticides under study have been extracted
H A McArthur et al.
Biochimica et biophysica acta, 568(2), 395-407 (1979-06-06)
Protoplast membranes and the particulate D,D-carboxypeptidase of Bacillus coagulans NCIB 9365 were extremely resistant to disruption by either detergents or urea. A combination of urea and the non-ionic detergent Genapol X-100 was required to achieve a significant solubilisation of membrane
Júlio C Fonseca et al.
Environmental monitoring and assessment, 75(1), 1-10 (2005-05-20)
This work reports studies of the degradation rates of a fatty alcohol polyglycol ether non-ionic surfactant, Genapol OXD-080, putatively useful for the control of red swamp crayfish (Procambarus clarkii Girard) in rice fields under laboratory and field conditions. The influence
E Chantelau
Diabetes & metabolism, 26(4), 304-306 (2000-09-30)
A case of a type 1 diabetic woman with auto-immune thyroiditis is reported, in whom repeated exposure to insulin containing Genapol(R) (polyethylen-polypropylenglycol) over 3 years reproducibly parallels with an increase of serum TSH (thyroid-stimulating hormone) above the normal limit. Previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service