G3769
Glutaryl-Ala-Ala-Phe-4-methoxy-β-naphthylamide
protease substrate
About This Item
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
powder
solubility
DMF: 50 mg/mL, clear, colorless to faintly yellow
storage temp.
−20°C
SMILES string
COc1cc(NC(=O)C(Cc2ccccc2)NC(=O)C(C)NC(=O)C(C)NC(=O)CCCC(O)=O)cc3ccccc13
InChI
1S/C31H36N4O7/c1-19(32-27(36)14-9-15-28(37)38)29(39)33-20(2)30(40)35-25(16-21-10-5-4-6-11-21)31(41)34-23-17-22-12-7-8-13-24(22)26(18-23)42-3/h4-8,10-13,17-20,25H,9,14-16H2,1-3H3,(H,32,36)(H,33,39)(H,34,41)(H,35,40)(H,37,38)
InChI key
QFJWKXYGBSQMDX-UHFFFAOYSA-N
Packaging
Substrates
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service