Skip to Content
Merck
All Photos(4)

Key Documents

G1279

Sigma-Aldrich

G 418 disulfate salt

powder, suitable for plant cell culture, BioReagent

Synonym(s):

Antibiotic G418

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product name

G 418 disulfate salt, suitable for plant cell culture, BioReagent

product line

BioReagent

Quality Level

form

powder

potency

≥720 μg per mg (Dried basis)

technique(s)

cell culture | plant: suitable

color

white to off-white

solubility

100 mg/mL (H2O clear)

antibiotic activity spectrum

Gram-negative bacteria

application(s)

agriculture

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](OC[C@@]1(C)O)O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H]([C@H](C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O

InChI

1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1

InChI key

UHEPSJJJMTWUCP-NKCAIAFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

G418 disulfate salt is an aminoglycoside antibiotic similar in structure to gentamicin used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene†. Optimal concentrations for selection and maintenance vary by cell lines. Bacteria and algae require 5 μg/mL or less while animal cells may require 300-500 μg/mL.

Caution

The product is stable for three years when stored at 2-8°C.

Preparation Note

G418 powder is soluble in water at 100 mg/mL.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frida C Mohlin et al.
Molecular immunology, 65(2), 367-376 (2015-03-04)
Atypical hemolytic uremic syndrome (aHUS) is a disease of complement dysregulation, characterized by hemolytic anemia, thrombocytopenia and acute renal failure. Mutations in complement inhibitors are major risk factors for development of aHUS. The three aHUS patients reported in this study
Yan-Ping Li et al.
STAR protocols, 2(2), 100438-100438 (2021-04-27)
Human-induced pluripotent stem cells (hiPSCs) can be differentiated into well-structured retinal organoids. In this protocol, we successfully established 3D retinae from patient-derived hiPSCs and built the retinitis pigmentosa model in vitro. Moreover, mutation in the retinitis pigmentosa GTPase regulator (RPGR)
Glenda Eger et al.
PloS one, 9(9), e109047-e109047 (2014-10-01)
The fibroblast mitogen platelet-derived growth factor -BB (PDGF-BB) induces a transient expression of the orphan nuclear receptor NR4A1 (also named Nur77, TR3 or NGFIB). The aim of the present study was to investigate the pathways through which NR4A1 is induced
Luis M Molinos-Albert et al.
Retrovirology, 11, 44-44 (2014-06-10)
The MPER region of the HIV-1 envelope glycoprotein gp41 is targeted by broadly neutralizing antibodies. However, the localization of this epitope in a hydrophobic environment seems to hamper the elicitation of these antibodies in HIV infected individuals.We have quantified and
Elsa A Marquez et al.
PloS one, 10(7), e0131308-e0131308 (2015-07-07)
Ly49 receptors can be peptide selective in their recognition of MHC-I-peptide complexes, affording them a level of discrimination beyond detecting the presence or absence of specific MHC-I allele products. Despite this ability, little is understood regarding the properties that enable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service