Skip to Content
Merck
All Photos(2)

Key Documents

G1025

Sigma-Aldrich

Gibberellic acid potassium salt

suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis

Synonym(s):

GA3, Gibberellin A3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21KO6
CAS Number:
Molecular Weight:
384.46
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
Pricing and availability is not currently available.

product line

BioReagent

Assay

~95%

composition

total GA3, ≥50%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

[K].[H][C@@]12CC[C@]3(O)C[C@]1(CC3=C)[C@@H](C(O)=O)[C@]4([H])[C@@]5(C)[C@@H](O)C=C[C@@]24OC5=O

InChI

1S/C19H22O6.K.H/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22;;/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22);;/t10-,11+,12-,13-,16-,17+,18+,19-;;/m1../s1

InChI key

YGDRGPFCIWRNTK-DHDXTLTISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gibberellin A3 (G1025, G7645) and A4 (G7276) may be used as supplements to plant growth media such as Murashige and Skoog media. The specific actions of A3 and A4 or combinations of Gebberellins should be determined in specific plant applications.

Biochem/physiol Actions

Gibberillic acids (GA) are important plant growth hormones that promote plant cell growth and elongation. Gebberillins promote rapid stem and root growth, induce mitotic division and initiate (break dormancy) and increase seed germination rates. The gibberellins are also involved in processes such as gravitropism, tensioning and floral display.[1] Auxins influence the biosynthesis and signaling of GA. GA is involved in auxin-mediated primary root elongation by modulating auxin signaling and transport.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abu Imran Baba et al.
International journal of molecular sciences, 20(14) (2019-07-25)
Seedling establishment following germination requires the fine tuning of plant hormone levels including that of auxin. Directional movement of auxin has a central role in the associated processes, among others, in hypocotyl hook development. Regulated auxin transport is ensured by
Khemais Abdellaoui et al.
Pesticide biochemistry and physiology, 107(1), 32-37 (2014-08-26)
Experiments were conducted to assess the effect of gibberellic acid (GA3), a plant growth regulator, on Locusta migratoria migratoria fifth instar larvae. Newly emerged larvae were exposed to various concentrations of GA3 administered by topical application or by forced ingestion.
Dante F Placido et al.
Plant biotechnology journal, 18(9), 1955-1968 (2020-02-08)
Drought stress is the major limiting factor in agriculture. Wheat, which is the most widely grown crop in the world, is predominantly cultivated in drought-prone rainfed environments. Since roots play a critical role in water uptake, root response to water
Zhigang Liao et al.
Nature communications, 10(1), 2738-2738 (2019-06-23)
The breeding of cereals with altered gibberellin (GA) signaling propelled the 'Green Revolution' by generating semidwarf plants with increased tiller number. The mechanism by which GAs promote shoot height has been studied extensively, but it is not known what causes
Feng Wang et al.
Plant biotechnology journal, 18(4), 1041-1055 (2019-10-05)
The ability to interpret daily and seasonal fluctuations, latitudinal and vegetation canopy variations in light and temperature signals is essential for plant survival. However, the precise molecular mechanisms transducing the signals from light and temperature perception to maintain plant growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service