Skip to Content
Merck
All Photos(1)

Key Documents

F9057

Sigma-Aldrich

FLLL31

≥98% (HPLC)

Synonym(s):

(E,E)-1,7-Bis(3,4-dimethoxyphenyl)-4,4-dimethyl-1,6-heptadiene-3,5-dione, Tetramethylcurcumin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28O6
CAS Number:
Molecular Weight:
424.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(\C=C\C(=O)C(C)(C)C(=O)\C=C\c2ccc(OC)c(OC)c2)cc1OC

InChI

1S/C25H28O6/c1-25(2,23(26)13-9-17-7-11-19(28-3)21(15-17)30-5)24(27)14-10-18-8-12-20(29-4)22(16-18)31-6/h7-16H,1-6H3/b13-9+,14-10+

InChI key

VMMZAMVBGQWOHT-UTLPMFLDSA-N

Biochem/physiol Actions

FLLL31 selectively binds to Janus kinase 2 and the STAT3 Src homology-2 (SH2) domain, effective inhibitors of STAT3 phosphorylation. STAT3 plays a critical role in early embryogenesis, but is largely dispensable in normal adult cells and tissues. On the other hand the JAK2/STAT3 signaling pathway is persistently activated in great number of human solid and blood cancers. Such activation is commonly associated with a worse prognosis. FLLL31 is a curcumin derivative locked in diketone-tautomeric form, which supposedly improves binding to SH2 domain. The compound inhibits JAK2 kinase activity and prevents STAT3 phosphorylation. FLLK31 is a potent and selective inhibitor of the STAT3 signaling pathway.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the JAKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bryan C Mounce et al.
Antiviral research, 142, 148-157 (2017-03-28)
Several compounds extracted from spices and herbs exhibit antiviral effects in vitro, suggesting potential pharmacological uses. Curcumin, a component of turmeric, has been used as a food additive and herbal supplement due to its potential medicinal properties. Previously, curcumin exhibited antiviral
Federica Liotti et al.
Cancers, 13(4) (2021-02-14)
Pattern recognition receptors (PRR) promote inflammation but also its resolution. We demonstrated that a specific PRR-formyl peptide receptor 1 (FPR1)-sustains an inflammation resolution response with anti-angiogenic and antitumor potential in gastric cancer. Since toll-like receptor 7 (TLR7) is crucial in
Daniela A Ferraro et al.
BMC cancer, 19(1), 312-312 (2019-04-06)
The tumour microenvironment is a critical regulator of malignant cancer progression. While endothelial cells have been widely studied in the context of tumour angiogenesis, their role as modulators of cancer cell invasion and migration is poorly understood. We have investigated
Rosmarie Michel-Schmidt et al.
International immunopharmacology, 29(1), 115-118 (2015-04-19)
Acetylcholine (ACh) acts as a local cellular signaling molecule and is widely expressed in nature, including mammalian cells and embryonic stem cells. The murine embryonic stem cell line CGR8 synthesizes and releases substantial amounts of ACh. Particularly during early differentiation
Shaopeng Yuan et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 36(6), 2340-2356 (2015-08-19)
Interleukin-17 (IL-17) is a major pro-inflammatory cytokine that initiates and maintains inflammation. However, the molecular mechanisms as to how IL-17 influences endothelial cells to promote neutrophil recruitment are not fully understood. Human endothelial cells (HMECs) were stimulated with IL-17, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service