F8067
Fetal Bovine Serum
New Zealand origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Synonym(s):
Fetal Bovine Serum, FBS, FCS, sera, serum
About This Item
Recommended Products
biological source
bovine fetus
Quality Level
sterility
sterile-filtered
product line
FBS Classic
composition
Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤20 mg/dL
origin
New Zealand origin
technique(s)
cell culture | hybridoma: suitable
cell culture | mammalian: suitable
impurities
≤10 EU/mL endotoxin
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
- Endotoxin and hemoglobin tested
- Tested for the presence of bacteria, virus, and mycoplasma
- Triple filtered with 0.1 micron membrane under aseptic conditions
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service