Skip to Content
Merck
All Photos(1)

Key Documents

F2426

Millipore

EZview Red ANTI-FLAG® M2 Affinity Gel

clone M2

Synonym(s):

Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32

clone

M2, monoclonal

analyte chemical class(es)

proteins

technique(s)

affinity chromatography: suitable
immunoprecipitation (IP): suitable

matrix

4% agarose bead; 45-165μm bead size

isotype

IgG1

capacity

≥0.6 mg/mL, gel binding capacity

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

EZ view Red Anti-FLAG M2 Affinity Gel is a resin that consists of Anti-FLAG M2 antibody, covalently bonded to 4% Agarose beads. The affinity gel is used to bind FLAG fusion proteins to samples, such as cell lysates and tissue, for purification of FLAG-tagged proteins in preparation of immunoprecipitation assays. Red dye enhances visability for more efficient results. Agarose beads bind at N-terminal, Met-N-terminal and C-terminal FLAG fusion proteins, 3xFLAG-tagged fusion proteins.

Specificity

Suitable for purifying N-terminal, Met-N-terminal, C-terminal FLAG fusion proteins, 3xFLAG fusion proteins.

Application

Immunoprecipitation (IP) of FLAG- and 3xFLAG-tagged fusion proteins.

Elution - FLAG peptide, Glycine, pH 3.5, 3x FLAG peptide

Learn more product details in our FLAG® application portal.

Physical form

1:1 (v/v) suspension in PBS containing 50% glycerol and 15 ppm Kathon

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
EZview is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

FLAG® Affinity Gels, FLAG® tag, 3xFLAG®tag, DYKDDDDK tag

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kara R Jones et al.
Molecular cancer therapeutics, 4(10), 1541-1547 (2005-10-18)
Incomplete DNA repair or misrepair can contribute to the cytotoxicity of DNA double-strand breaks. Consequently, interference with double-strand break repair, by pharmacologic or genetic means, is likely to sensitize tumor cells to ionizing radiation. The current studies were designed to
Matthew S Walters et al.
Journal of virology, 84(13), 6861-6865 (2010-04-16)
Varicella zoster virus encodes an immediate-early (IE) protein termed ORF61p that is orthologous to the herpes simplex virus IE protein ICP0. Although these proteins share several functional properties, ORF61p does not fully substitute for ICP0. The greatest region of similarity
W Du et al.
Cell death and differentiation, 16(11), 1493-1504 (2009-07-11)
The tumor suppressor p53 induces potent anti-proliferative responses in stressed cells; in unstressed cells this ability of p53 is restrained by Hdm2. Expression of Hdm2 is also induced by p53, thereby establishing feedback inhibition. Regulation of the p53-Hdm2 interaction and
Jiahai Zhou et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(39), 14343-14348 (2006-09-16)
The unfolded protein response (UPR) is an evolutionarily conserved mechanism by which all eukaryotic cells adapt to the accumulation of unfolded proteins in the endoplasmic reticulum (ER). Inositol-requiring kinase 1 (IRE1) and PKR-related ER kinase (PERK) are two type I
Ting-Ting Qu et al.
BMC cancer, 18(1), 27-27 (2018-01-06)
Lin28B and its paralog Lin28A are small RNA binding proteins that have similar inhibitory effects, although they target separate steps in the maturation of let-7 miRNAs in mammalian cells. Because Lin28B participates in the promotion and development of tumors mostly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service