EP951020600
Eppendorf® twin.tec PCR Plates 96
Optimized rim for barcodes, semi-skirted, well volume 250 μL, colorless, pkg of 300 ea
All Photos(1)
About This Item
UNSPSC Code:
41106300
material:
colorless
conical bottom
polypropylene wells
conical bottom
polypropylene wells
size:
96 well
sterility:
non-sterile
feature:
lid: no
semi-skirted
semi-skirted
Recommended Products
material
colorless
conical bottom
polypropylene wells
sterility
non-sterile
feature
lid: no
semi-skirted
packaging
pkg of 300 ea
size
96 well
well volume
250 μL
suitability
suitable for PCR
General description
Eppendorf® twin.tec PCR Plate 96, semi-skirted, 250 µL, PCR clean, colorless, 300 plates
- Semi-skirted for label and barcode
- Fits most thermal cyclers
- 250 µL max. well volume (when used with cap strips)
- Improved well-to-well tolerance
- Available with barcode (upon request), for skirted and semi-skirted only
Features and Benefits
- Semi-skirted for label and barcode
- Fits most thermal cyclers
- 250 µL max. well volume (when used with cap strips)
- Improved well-to-well tolerance
- Available with barcode (upon request), for skirted and semi-skirted only
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)