Skip to Content
Merck
All Photos(1)

Key Documents

EP951020443

Eppendorf twin.tec® PCR Plates 96

skirted, size 150 μL, green, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
size:
150 μL
96 well
feature:
skirt
skirted
Pricing and availability is not currently available.

feature

skirt
skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020443

technique(s)

PCR: suitable

size

150 μL
96 well

color

green

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Eppendorf “twin.tec” PCR Plates 96 are PCR plates consisting of a rigid polycarbonate framework and 96 thin-walled polypropylene wells. These plates are used extensively in various molecular biology applications, particularly PCR.

Application

Eppendorf “twin.tec” PCR Plates 96 are used in most PCR applications ranging from standard PCR to Real time PCR and High throughput PCR. Additionally, they are also well suited for handling and processing of PCR samples in low volume quantities.
PCR

Features and Benefits

  • Eppendorf “twin.tec” PCR Plates 96 have skirted wells with a capacity of 150 µL.
  • These plates are green in color and PCR clean.
  • The presence of eight holes in the skirted wells helps in plate positioning and removal of the plates from the thermal blocks.
  • The low-profile design of the plate enhances the efficiency of PCR for small sample volumes.
  • Optimized rims support the barcode system and automated work processes.
  • Twin.tec plates are ideal for quantitative real time PCR.

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service