Skip to Content
Merck
All Photos(1)

Documents

EP951010022

Eppendorf® PCR Eight-tube strips

capacity 0.2 mL, pack of 960 ea (120 x strips of 8 tubes)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

polypropylene

sterility

non-sterile

packaging

pack of 960 ea (120 x strips of 8 tubes)

manufacturer/tradename

Eppendorf® 951010022

capacity

0.2 mL

color

colorless

General description

Eppendorf PCR Tube Strips, 0.2 mL, PCR clean, 8-tube strips, 120 pcs. (960 tubes)
  • Contamination shield on hinged lid
  • Defined lid position due to special hinge
  • High transparency even on the tube base
  • Etched lid for labeling
  • Tight sealing
  • Easy to open
  • For use with all thermal cyclers with 0.2 mL block format
  • Also available in 8-tube strip format
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)1)

1) Download of certificates under www.eppendorf.com/certificates

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service