EP022363557
Eppendorf® Flex-Tubes Microcentrifuge Tubes
volume 1.5 mL, green, pack of 500 tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
green
polypropylene
sterility
non-sterile
feature
graduated
Flex-Tube lid
packaging
pack of 500 tubes
manufacturer/tradename
Eppendorf® 022363557 (NA)
technique(s)
PCR: suitable
volume
1.5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The easy-open, easy-close design of the Flex-Tube provides convenient, ergonomic, and reliable one-handed operation. Whether sample preparation, centrifugation or storage – the Flex-Tube fit perfectly into your workflows. This is the successor to the renowned first microcentrifuge tube 3810 from Eppendorf. The idea of manufacturing a 1.5 mL plastic test tube was born out of the necessity to introduce an appropriate container into the automatic clinical analyzers developed by Eppendorf in 1962. The tried and tested design with invaluable features shows perfect sealing qualities, excellent chemical stability and optimal construction materials for resisting the demands of centrifugal forces. As they are easy to open and close securely, The Eppendorf Flex-Tubes off er comfort and dependability for sample preparation, centrifugation and storage. One vessel for all applications in molecular biology, chemistry and bioche mistry – safer than ever. The Eppendorf Flex-Tubes can be centrifuged up to 30,000 x g without any problems.
Application
Eppendorf™Flex-Tubes Microcentrifuge Tubes are ideal for preparing samples, storage, or centrifugation
Features and Benefits
- Functionality is guaranteed from -86 C to 100 C
- Simple verifi cation of the pipetted volume by clear graduation
- All variants can be autoclaved for subsequent use in sterile environments (121 C, 20 min.)
- Manufactured without the use of slip agents, plasticizers and biocides
- High resistance to chemicals, mechanical stress and temperature extremes – manufactured from virgin polypropylene
- Tube lid is easy to open and close for ergonomic handling
- Easy labeling on frosted lid and surface on the side
- Flex-Tube lid is easy to open and close for ergonomic operation
- Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes
- g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- Graduation for visual control of the pipetted volume
- Frosted lid and surface on the side for easy labeling
- Ensured functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service