Skip to Content
Merck
All Photos(1)

Key Documents

EP0030127978

Eppendorf® PCR Sealing Mat

PCR clean, for DWP 96/1000 and DWP 96/500, pkg of 50 ea (5 bags of 10 ea)

Synonym(s):

PCR, Plates, Sealing films

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

sterility

non-sterile

feature

PCR clean
for DWP 96/1000 and DWP 96/500

packaging

pkg of 50 ea (5 bags of 10 ea)

manufacturer/tradename

Eppendorf® 0030127978

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Eppendorf® PCR sealing mats provide the optimal sealing solutions for various laboratory applications
  • Designed to seal Eppendorf® deep-well plates - DWP 96/1000 and DWP 96/500
  • Highly chemical resistant to most of the substances in the laboratory
  • Ideal for short-term storage of samples
  • Reusable and autoclavable

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service