Skip to Content
Merck
All Photos(1)

Key Documents

EMU037431

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Syp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTGGGTCTTTGCCATCTTCGCCTTTGCTACGTGCGGCAGCTACACCGGAGAGCTTCGGCTGAGCGTGGAGTGTGCCAACAAGACGGAGAGTGCCCTCAACATCGAAGTCGAATTTGAGTACCCATTCAGGCTGCACCAAGTGTACTTTGATGCACCCTCCTGCGTTAAAGGGGGCACTACCAAGATCTTCCTAGTTGGTGACTACTCCTCCTCGGGTGAATTCTTTGTCACCGTGGCTGTGTTTGCCTTCCTCTACTCCATGGGGGCCCTGGCCACCTACATCTTCCTGCAGAACAAGTACCGAGAGAACAACAAAGGGCCAATGATGGACTTCCTGGCCACAGCAGTGTTCGCTTTCATGTGGCTAGTTAGCTCATCCGCCTGGGCCAAAGGCCTGTCCGATGTGAAGATGGCCACTGACCCAGAGAACATTATCAAGGAGATGCCTATGTGCCGCCAGACAGGAAACACATGCAAGGAACTGAGGGACCCTGT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toru Mitsumori et al.
Experimental hematology, 42(9), 783-792 (2014-05-28)
The hypoxic microenvironment of the bone marrow, known as the hypoxic niche, supports hematopoietic stem cell quiescence and maintains long-term repopulation activity. Hypoxia also affects the expansion of progenitor cells and enhances erythropoiesis and megakaryopoiesis. In contrast to the known
Hsueh-Chun Wang et al.
BMC cancer, 14, 442-442 (2014-06-17)
Tumor invasion and metastasis represent a major unsolved problem in cancer pathogenesis. Recent studies have indicated the involvement of Src-homology 2 domain-containing tyrosine phosphatase 2 (SHP2) in multiple malignancies; however, the role of SHP2 in oral cancer progression has yet
Rong-Ping Zhou et al.
Molecular medicine reports, 11(6), 4489-4495 (2015-01-31)
Chlorogenic acid (CGA) exhibits various biological properties, including the inhibition of oxidation, obesity, apoptosis and tumorigenesis. CGA is also able to promote cell survival and proliferation. The aim of the present study was to determine the effects and underlying molecular
K S Siveen et al.
British journal of cancer, 111(7), 1327-1337 (2014-08-08)
Constitutive activation of signal transducer and activator of transcription signalling 3 (STAT3) has been linked with survival, proliferation and angiogenesis in a wide variety of malignancies including hepatocellular carcinoma (HCC). We evaluated the effect of lupeol on STAT3 signalling cascade

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service