Skip to Content
Merck
All Photos(1)

Key Documents

EHU134551

Sigma-Aldrich

MISSION® esiRNA

targeting human NRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGCCTGAACTACCCTGAGAATGGGTGGACTCCCGGAGAGGATTCCTACCGAGAGTGGATACAGGTAGACTTGGGCCTTCTGCGCTTTGTCACGGCTGTCGGGACACAGGGCGCCATTTCAAAAGAAACCAAGAAGAAATATTATGTCAAGACTTACAAGATCGACGTTAGCTCCAACGGGGAAGACTGGATCACCATAAAAGAAGGAAACAAACCTGTTCTCTTTCAGGGAAACACCAACCCCACAGATGTTGTGGTTGCAGTATTCCCCAAACCACTGATAACTCGATTTGTCCGAATCAAGCCTGCAACTTGGGAAACTGGCATATCTATGAGATTTGAAGTATACGGTTGCAAGATAACAGATTATCCTTGCTCTGGAATGTTGGGTATGGTGTCTGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pezhman Salehi et al.
PLoS genetics, 13(10), e1007048-e1007048 (2017-10-24)
Neuropilin-1 (Nrp1) encodes the transmembrane cellular receptor neuropilin-1, which is associated with cardiovascular and neuronal development and was within the peak SNP interval on chromosome 8 in our prior GWAS study on age-related hearing loss (ARHL) in mice. In this
Rebecca McLennan et al.
Developmental biology, 407(1), 12-25 (2015-08-19)
Embryonic neural crest cells travel in discrete streams to precise locations throughout the head and body. We previously showed that cranial neural crest cells respond chemotactically to vascular endothelial growth factor (VEGF) and that cells within the migratory front have
Toru Nakanishi et al.
Cell death & disease, 10(2), 67-67 (2019-01-27)
Following incomplete spinal cord injury (SCI), reorganization of the corticospinal tract (CST) contributes to spontaneous motor recovery. Axotomized CST fibers form collaterals and make synapses with interneurons, followed by pruning of excess fibers. Although axonal pruning is involved in refinement
Juan Cong Dong et al.
Journal of cellular and molecular medicine, 19(9), 2286-2295 (2015-07-07)
The purpose of this study was to determine the correlation between over-expression of the neuropilin 1 (NRP1) gene and growth, survival, and radio-sensitivity of non-small cell lung carcinoma (NSCLC) cells. 3-[4,5-dimethylthylthiazol-2-yl]-2,5 diphenyltetrazolium broide (MTT) and colony assays were then performed
Jiahui Cao et al.
Nature communications, 8(1), 2210-2210 (2017-12-22)
VEGFR-2/Notch signalling regulates angiogenesis in part by driving the remodelling of endothelial cell junctions and by inducing cell migration. Here, we show that VEGF-induced polarized cell elongation increases cell perimeter and decreases the relative VE-cadherin concentration at junctions, triggering polarized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service