Skip to Content
Merck
All Photos(1)

Key Documents

EHU093481

Sigma-Aldrich

MISSION® esiRNA

targeting human CFLAR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCATCAGGTTGAAGAAGCACTTGATACAGATGAGAAGGAGATGCTGCTCTTTTTGTGCCGGGATGTTGCTATAGATGTGGTTCCACCTAATGTCAGGGACCTTCTGGATATTTTACGGGAAAGAGGTAAGCTGTCTGTCGGGGACTTGGCTGAACTGCTCTACAGAGTGAGGCGATTTGACCTGCTCAAACGTATCTTGAAGATGGACAGAAAAGCTGTGGAGACCCACCTGCTCAGGAACCCTCACCTTGTTTCGGACTATAGAGTGCTGATGGCAGAGATTGGTGAGGATTTGGATAAATCTGATGTGTCCTCATTAATTTTCCTCATGAAGGATTACATGGGCCGAGGCAAGATAAGCAAGGAGAAGAGTTTCTTGGACCTTGTGGTTGAGTTGGAGAAACTAAATCTGGTTGCCCCAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H H Cheung et al.
Cell death & disease, 2, e146-e146 (2011-04-15)
Smac mimetic compounds (SMCs) are experimental small molecules that induce tumour necrosis factor alpha (TNFα)-dependent cancer cell death by targeting the inhibitor of apoptosis proteins. However, many cancer cell lines are resistant to SMC-mediated apoptosis despite the presence of TNFα.
Reyaz ur Rasool et al.
Scientific reports, 6, 18800-18800 (2016-01-06)
The eukaryotic translation initiation factor 4E (eIF4E) is considered as a key survival protein involved in cell cycle progression, transformation and apoptosis resistance. Herein, we demonstrate that medicinal plant derivative 3-AWA (from Withaferin A) suppressed the proliferation and metastasis of
Therese Bredholt et al.
Molecular cancer, 8, 101-101 (2009-11-17)
An organic extract of the recreational herb khat (Catha edulis Forsk.) triggers cell death in various leukemia cell lines in vitro. The chemotherapeutics camptothecin, a plant alkaloid topoisomerase I inhibitor, was tested side-by-side with khat in a panel of acute
Andre van Krüchten et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(5), 2779-2793 (2018-02-07)
Superinfections with Staphylococcus aureus are a major complication of influenza disease, causing excessive inflammation and tissue damage. This enhanced cell-damaging effect is also observed in superinfected tissue cultures, leading to a strong decrease in overall cell viability. In our analysis
Yao Wang et al.
Frontiers in oncology, 9, 857-857 (2019-09-26)
Immune checkpoint blockade of programmed cell death protein 1 (PD-1) had an impressive long-lasting effect in a portion of advanced-stage melanoma patients, however, this therapy failed to induce responses in several patients; how to increase the objective response rate is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service