Skip to Content
Merck
All Photos(1)

Key Documents

EHU074571

Sigma-Aldrich

MISSION® esiRNA

targeting human VHL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATCTGGAAGACCACCCAAATGTGCAGAAAGACCTGGAGCGGCTGACACAGGAGCGCATTGCACATCAACGGATGGGAGATTGAAGATTTCTGTTGAAACTTACACTGTTTCATCTCAGCTTTTGATGGTACTGATGAGTCTTGATCTAGATACAGGACTGGTTCCTTCCTTAGTTTCAAAGTGTCTCATTCTCAGAGTAAAATAGGCACCATTGCTTAAAAGAAAGTTAACTGACTTCACTAGGCATTGTGATGTTTAGGGGCAAACATCACAAAATGTAATTTAATGCCTGCCCATTAGAGAAGTATTTATCAGGAGAAGGTGGTGGCATTTTTGCTTCCTAGTAAGTCAGGACAGCTTGTATGTAAGGAGGTTTGTATAAGTAATTCAGTGGGAATTGCAGCATA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Hao et al.
Neurochemical research, 41(9), 2391-2400 (2016-06-22)
The VHL (Von Hippel-Lindau) gene is a tumor suppressor gene, which is best known as an E3 ubiquitin ligase that negatively regulates the hypoxia inducible factor. The inactivation of VHL gene could result in the abnormal synthesis of VHL protein
Yinghong Zhou et al.
Oxidative medicine and cellular longevity, 2020, 7079308-7079308 (2020-04-11)
Hepatocellular carcinoma (HCC) is regarded as a leading cause of cancer-related deaths, and its progression is associated with hypoxia and the induction of hypoxia-inducible factor (HIF). Meloxicam, a selective cyclooxygenase-2 (COX-2) inhibitor, induces cell death in various malignancies. However, the
Susan E Scanlon et al.
Oncotarget, 9(4), 4647-4660 (2018-02-13)
The von Hippel-Lindau (
Dawei Li et al.
Free radical biology & medicine, 110, 102-116 (2017-06-07)
Oxidative stress has a critical role in the pathogenesis of acetaminophen (APAP) induced hepatocellular necrosis, and the identification of novel approaches to attenuate oxidative stress is essential to prevent/revert the disease. This study investigated the role of both HIF-1 and
Mian Wei et al.
Journal of cellular physiology, 234(10), 17392-17404 (2019-02-23)
Microenvironmental hypoxia-mediated drug resistance is responsible for the failure of cancer therapy. To date, the role of the hedgehog pathway in resistance to temozolomide (TMZ) under hypoxia has not been investigated. In this study, we discovered that the increasing hypoxia-inducible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service