Skip to Content
Merck
All Photos(1)

Key Documents

EHU060601

Sigma-Aldrich

MISSION® esiRNA

targeting human DNMT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAGCCACAGATGCTGACAAATGAGAAGCTGTCCATCTTTGATGCCAACGAGTCTGGCTTTGAGAGTTATGAGGCGCTTCCCCAGCACAAACTGACCTGCTTCAGTGTGTACTGTAAGCACGGTCACCTGTGTCCCATCGACACCGGCCTCATCGAGAAGAATATCGAACTCTTCTTTTCTGGTTCAGCAAAACCAATCTATGATGATGACCCATCTCTTGAAGGTGGTGTTAATGGCAAAAATCTTGGCCCCATAAATGAATGGTGGATCACTGGCTTTGATGGAGGTGAAAAGGCCCTCATCGGCTTCAGCACCTCATTTGCCGAATACATTCTGATGGATCCCAGTCCCGAGTATGCGCCCATATTTGGGCTGATGCAGGAGAAGATCTACATCAGCAAGATTGTGGTGGAGTTCCTGCAGAGCAATTCCGACTCGACCTATGAGGACCTGATCAACAAGATCGAGACCACGGTTCCTCCTTCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuhao Fu et al.
Experimental eye research, 165, 47-58 (2017-09-13)
The principle reason of high failure rate of glaucoma filtration surgery is the loss of filtration function caused by postoperative scar formation. We investigated the effects of 5-aza-2'-deoxycytidine (5-Aza-dc), a DNA methyltransferases inhibitor, on human Tenon's capsule fibroblasts (HTFs) differentiation
Arunagiri Kuha Deva Magendhra Rao et al.
Molecular oncology, 13(6), 1342-1355 (2019-04-09)
Breast cancer is the most common malignancy among women, with the highest incidence rate worldwide. Dysregulation of long noncoding RNAs during the preliminary stages of breast carcinogenesis is poorly understood. In this study, we performed RNA sequencing to identify long
Luisa Tomasello et al.
International journal of molecular sciences, 21(7) (2020-04-01)
Protein tyrosine phosphatase receptor type γ (PTPRG) is a tumor suppressor gene, down-regulated in Chronic Myeloid Leukemia (CML) cells by the hypermethylation of its promoter region. β-catenin (CTNNB1) is a critical regulator of Leukemic Stem Cells (LSC) maintenance and CML
Luc Gailhouste et al.
Cell death & disease, 9(5), 468-468 (2018-04-28)
Curative management of pancreatic adenocarcinoma is limited because this malignancy remains resistant to most chemotherapeutic drugs. Strategies that reverse epigenetic alterations offer a unique opportunity for cancer cell reprogramming, which is valuable for development of new treatments. The aim of
Wen-Long Xue et al.
American journal of physiology. Cell physiology, 318(5), C857-C869 (2020-03-19)
Diabetes (especially Type II) is one of the primary threats to cardiovascular health. Wound healing defects and vascular dysfunction are common in diabetic patients, and the primary cause of deterioration is sustained high plasma glucose. microRNA, a noncoding RNA, has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service